Skip to Content
Merck
All Photos(1)

Key Documents

SML2307

Sigma-Aldrich

Lodoxamide

≥98% (HPLC)

Synonym(s):

2,2′-[(2-Chloro-5-cyano-1,3-phenylene)diimino]bis-2-oxoacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€81.30
25 MG
€329.00

€81.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€81.30
25 MG
€329.00

About This Item

Empirical Formula (Hill Notation):
C11H6ClN3O6
CAS Number:
Molecular Weight:
311.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€81.30


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to very dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

Clc1c(cc(cc1NC(=O)C(=O)O)C#N)NC(=O)C(=O)O

InChI

1S/C11H6ClN3O6/c12-7-5(14-8(16)10(18)19)1-4(3-13)2-6(7)15-9(17)11(20)21/h1-2H,(H,14,16)(H,15,17)(H,18,19)(H,20,21)

InChI key

RVGLGHVJXCETIO-UHFFFAOYSA-N

Biochem/physiol Actions

Lodoxamide Tromethamine is a potent agonist of GPR35 in both human and rat, and an antiallergic mast cell stabilizer used clinically in the UK for treatment of allergic conjunctivitis.
potent agonist of GPR35 in both human and rat, and an antiallergic mast cell stabilizer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soo-Jin Park et al.
British journal of pharmacology, 175(1), 154-161 (2017-10-27)
GPR35 has long been considered an orphan GPCR, because no endogenous ligand of GPR35 has been discovered. CXCL17 (a chemokine) has been reported to be an endogenous ligand of GPR35, and it has even been suggested that it be called
Alejandro Rodriguez-Garcia et al.
International ophthalmology, 38(3), 1243-1249 (2017-06-12)
To report the therapeutic efficacy and safety of topical 0.1% lodoxamide in the long-term treatment of superior limbic keratoconjunctivitis. Sixty-seven eyes of 34 patients with active SLK were studied. Therapeutic response was analyzed according to modified-Ohashi parameters. All eyes were
Amanda E MacKenzie et al.
Molecular pharmacology, 85(1), 91-104 (2013-10-12)
Lack of high potency agonists has restricted analysis of the G protein-coupled receptor GPR35. Moreover, marked variation in potency and/or affinity of current ligands between human and rodent orthologs of GPR35 has limited their productive use in rodent models of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service