Skip to Content
Merck

Skip To

SML2217

Parecoxib sodium

≥98% (HPLC)

Promotion Available

Synonym(s):

SC-69124, Sodium (4-(5-methyl-3-phenylisoxazol-4-yl)phenylsulfonyl)(propionyl)amide

Sign In to View Organizational & Contract Pricing.

Spend € 450 — Save 5 %; Spend € 700 — Save 10 %; Spend € 1.500 — Save 15 %*

Sign in to check eligibility. Shop Products

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 344,00

About This Item

Empirical Formula (Hill Notation):
C19H17N2O4S · Na
CAS Number:
Molecular Weight:
392.40
NACRES:
NA.77
UNSPSC Code:
51111800
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

€ 344,00

Save Up To 15 %


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(C2=CC=C(S([N-]C(CC)=O)(=O)=O)C=C2)C(C3=CC=CC=C3)=NO1.[Na+]

InChI

1S/C19H18N2O4S.Na/c1-3-17(22)21-26(23,24)16-11-9-14(10-12-16)18-13(2)25-20-19(18)15-7-5-4-6-8-15;/h4-12H,3H2,1-2H3,(H,21,22);/q;+1/p-1

InChI key

HQPVVKXJNZEAFW-UHFFFAOYSA-M

Biochem/physiol Actions

Non-steroidal anti-inflammatory drug (NSAID), a cyclooxygenase-2 (COX-2) selective inhibitor
Parecoxib sodium is a non-steroidal anti-inflammatory drug (NSAID), a cyclooxygenase-2 (COX-2) selective inhibitor. It is a water-soluble and injectable prodrug of valdecoxib.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2014SML1674SML2632
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

desiccated

storage condition

-

storage condition

-

storage condition

-

solubility

H2O: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 30 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to brown

color

white to beige

color

white to beige


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters