Skip to Content
Merck
All Photos(1)

Key Documents

SML1674

Sigma-Aldrich

UPCDC30245

≥98% (HPLC)

Synonym(s):

1-(3-(5-Fluoro-1H-indol-2-yl)phenyl)-N-(2-(4-isopropylpiperazin-1-yl)ethyl)piperidin-4-amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€127.00
25 MG
€491.00

€127.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€127.00
25 MG
€491.00

About This Item

Empirical Formula (Hill Notation):
C28H38FN5
CAS Number:
Molecular Weight:
463.63
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€127.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C(NC(C2=CC=CC(N3CCC(NCCN4CCN(C(C)C)CC4)CC3)=C2)=C5)C5=C1

InChI key

LZHXZCVDLATFAR-UHFFFAOYSA-N

Biochem/physiol Actions

UPCDC30245 is an allosteric inhibitor of AAA ATPase p97, also called valosine containing protein (VCP). p97 is an integral component of the ubiquitin fusion degradation (UFD) pathway. It plays a role in degradation of misfolded proteins, Golgi membrane reassembly, membrane transport, myofibril assembly, autophagosome maturation, and cell division, and is overexpressed in many tumor types. UPCDC30245 has an IC50 value of 27 nM, and has been shown to bind at the junction between the D1 and D2 domains of p97. UPCDC30245 has been used in cryo-ECM to show how such inhibitor binding prevents the confomational changes necessary for p97 function.
UPCDC30245 is an allosteric inhibitor of AAA ATPase p97.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service