Skip to Content
Merck
All Photos(1)

Key Documents

SML2203

Sigma-Aldrich

SGC-GAK-1N

≥98% (HPLC)

Synonym(s):

CA71, CA93 Negative Control, N-Methyl-6-(trifluoromethyl)-N-(3,4,5-trimethoxyphenyl)quinolin-4-amine, SGC-GAK-1 Negative Control

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30
25 MG
€363.00

€89.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€89.30
25 MG
€363.00

About This Item

Empirical Formula (Hill Notation):
C20H19F3N2O3
CAS Number:
Molecular Weight:
392.37
UNSPSC Code:
41106609
NACRES:
NA.77

€89.30


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

dark green to black

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C1=CC(OC)=C(OC)C(OC)=C1)C2=C3C(C=CC(C(F)(F)F)=C3)=NC=C2

InChI

1S/C20H19F3N2O3/c1-25(13-10-17(26-2)19(28-4)18(11-13)27-3)16-7-8-24-15-6-5-12(9-14(15)16)20(21,22)23/h5-11H,1-4H3

InChI key

PVTQCCFMFWASHK-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

Negative control analog for the cyclin G-associated kinase (GAK) inhibitor SGC-GAK-1 (CA93).
SGC-GAK-1N (CA71) is a structure analog of and the recommended negative control for the ATP-competitive cyclin G-associated kinase (GAK) inhibitor SGC-GAK-1 (CA93) (GAK KD = 1.9 nM/SGC-GAK-1 vs. 7.1 μM/SGC-GAK-1N by ligand competitive binding assay; GAK IC50 = 110 nM/SGC-GAK-1 vs. >50 μM/SGC-GAK-1N by cellular target engagement assay; Dengue viral entry and packaging IC50 = 920 nM/SGC-GAK-1 vs. >10 μM/SGC-GAK-1N). For characterization details of the active probe, SGC-GAK-1, please visit the SGC-GAK-1 probe summary on the Structural Genomics Consortium (SGC) website.

SGC-GAK-1, the active probe, is available from Sigma. To learn more about and purchase SGC-GAK-1, click here.

To learn about other SGC chemical probes, visit sigma.com/sgc

Features and Benefits

SGC-GAK-1N is a negative control for SGC-GAK-1, a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service