Skip to Content
Merck
All Photos(1)

Key Documents

5.38770

Sigma-Aldrich

GAK Inhibitor

Synonym(s):

GAK Inhibitor, Cyclin G Associated Kinase Inhibitor, Auxilin 2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5387700001
€192.00

€192.00


Estimated to ship on25 May 2025



Select a Size

Change View
5387700001
€192.00

About This Item

Empirical Formula (Hill Notation):
C19H21N3O4S
Molecular Weight:
387.45
UNSPSC Code:
12352200
NACRES:
NA.77

€192.00


Estimated to ship on25 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable isothiazolopyridine based compound that acts as a high-affinity binding, reversible and ATP-competitive inhibitor of cyclin G associated kinase (GAK; Kd = 8.3 nM). Displays moderate to excellent selectivity over Kit, PDGFRβ, Flt3, Mek5 & PDGFRα (Kd = 29, 70, 110, 150 & 220 nM, respectively) in a 456-kinase profiling. Shown to dose-dependently disrupt both HCV entry & assembly and efficiently arrest HCV infection (EC50 = 2.55 & 3.6 µM in HCVcc- & HCVpp-infected Huh-7.5 cells, respectively; CC50 = 23.27 µM in HCVcc-infected Huh-7.5 cells). Significantly inhibits intracellular & extracellular infectivity (EC50 = 1.64 & 2.13 µM, respectively) and AP2M1 phosphorylation. Exerts no effect on HCV RNA replication.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
GAK
Reversible: yes
Secondary Target
Kit, PDGFRβ, Flt3, Mek5 & PDGFRα

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kovackova, S., et al. 2015. J. Med. Chem.58, 3393.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service