Skip to Content
Merck

Skip To

SML1998

KLH45

≥98% (HPLC)

Synonym(s):

N-Cyclohexyl-N-phenethyl-4-(4-(trifluoromethoxy)phenyl)-2H-1,2,3-triazole-2-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 108,00
25 mg
Check Cart for Availability
€ 295,00

About This Item

Empirical Formula (Hill Notation):
C24H25F3N4O2
CAS Number:
Molecular Weight:
458.48
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
oil

€ 108,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

oil

color

colorless to light yellow

storage temp.

2-8°C

SMILES string

O=C(N(CCC1=CC=CC=C1)C2CCCCC2)N(N=C3)N=C3C4=CC=C(OC(F)(F)F)C=C4

InChI

1S/C24H25F3N4O2/c25-24(26,27)33-21-13-11-19(12-14-21)22-17-28-31(29-22)23(32)30(20-9-5-2-6-10-20)16-15-18-7-3-1-4-8-18/h1,3-4,7-8,11-14,17,20H,2,5-6,9-10,15-16H2

InChI key

FYOYNRLSBYWAHL-UHFFFAOYSA-N

Biochem/physiol Actions

In vivo-active, potent and selective irreversible inhibitor of DDHD2
KLH45 is an in vivo-active, potent and selective irreversible inhibitor of DDHD2. KLH45 inactivates brain levels of DDHD2 and increases levels of triglycerides in mouse brain.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0358SML2092SML1602
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

oil

form

film

form

film

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

colorless to light yellow

color

colorless

color

colorless

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters