Skip to Content
Merck
All Photos(1)

Key Documents

SML0358

Sigma-Aldrich

AA74-1

≥98% (HPLC)

Synonym(s):

[4-(4-Hydroxy-2,6-dimethylheptan-4-yl)-2H-1,2,3-triazol-2-yl](pyrrolidin-1-yl)methanone, [4-[1-Hydroxy-3-methyl-1-(2-methylpropyl)butyl]-2H-1,2,3-triazol-2-yl]-1-pyrrolidinylmethanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€182.00

€182.00


Estimated to ship on13 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€182.00

About This Item

Empirical Formula (Hill Notation):
C16H28N4O2
CAS Number:
Molecular Weight:
308.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€182.00


Estimated to ship on13 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

film

color

colorless

solubility

DMSO: >15 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)CC(O)(CC(C)C)c1cnn(n1)C(=O)N2CCCC2

InChI

1S/C16H28N4O2/c1-12(2)9-16(22,10-13(3)4)14-11-17-20(18-14)15(21)19-7-5-6-8-19/h11-13,22H,5-10H2,1-4H3

InChI key

GXBVPGCIMCIASQ-UHFFFAOYSA-N

Biochem/physiol Actions

AA74-1 is an extremely potent and selective inhibitor of the serine hydrolase acyl-peptide hydrolase (APEH) with an IC50 of 5 nM in mouse T-cell proteomes. AA74-1 potently blocked the activity of its target serine hydrolase, APEH, in mice in vivo without significantly affecting other enzymes. The APEH inhibition caused accumulation of N-acetylated proteins and stimulated cellular proliferation in T cells.

The APEH gene is deleted in some cancers, in which it has been proposed to serve as a potential tumor suppressor, and is involved in the degradation of oligomeric amyloid-beta which could make it a new target for therapy aimed at reducing neurodegeneration in Alzheimer′s disease. AA74-1 is the first selective tool for studying APEH activity.
AA74-1 is an extremely potent and selective inhibitor of the serine hydrolase acyl-peptide hydrolase (APEH).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rubayet Elahi et al.
mSphere, 4(3) (2019-05-10)
The human malaria parasite Plasmodium falciparum causes disease as it replicates within the host's erythrocytes. We have found that an erythrocyte serine hydrolase, acylpeptide hydrolase (APEH), accumulates within developing asexual parasites. Internalization of APEH was associated with a proteolytic event

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service