Skip to Content
Merck
All Photos(1)

Key Documents

SML1947

Sigma-Aldrich

ITH12575

≥98% (HPLC)

Synonym(s):

4,1-Benzothiazepin-2(3H)-one, 7-chloro-1,5-dihydro-5-[2-(1-methylethyl)phenyl]-, 7-Chloro-5-(2′-isopropylphenyl)-3,5-dihydro-4,1-benzothiazepin-2-(1H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€86.80

€86.80


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€86.80

About This Item

Empirical Formula (Hill Notation):
C18H18ClNOS
CAS Number:
Molecular Weight:
331.86
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€86.80


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C)C1=C(C2C(C=C(Cl)C=C3)=C3NC(CS2)=O)C=CC=C1

InChI

1S/C18H18ClNOS/c1-11(2)13-5-3-4-6-14(13)18-15-9-12(19)7-8-16(15)20-17(21)10-22-18/h3-9,11,18H,10H2,1-2H3,(H,20,21)

InChI key

RDMUJSJZBYAAEF-UHFFFAOYSA-N

Biochem/physiol Actions

ITH12575 is apotent and selective inhibitor of mitochondrial Na+/Ca2+ exchanger mNCX with neuroprotective activity. It was shown to reduce Ca2+ efflux from the mitochondria to the cytosol with an EC50 value of 690 nM and was shown to protect against Ca2+ overload and mitigated the neuronal damage in a model of oxidative stress. At higher concentrations ITH12575 also reduced Ca2+ influx through CALHM1, a Ca2+ channel that plays a key role in the neuronal electrical activity.
Inhibitor of Mitochondrial Na+/Ca2+ exchanger (mNCX)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco J Martínez-Sanz et al.
European journal of medicinal chemistry, 109, 114-123 (2016-01-17)
The mitochondrial Na(+)/Ca(2+) exchanger plays an important role in the control of cytosolic Ca(2+) cycling in excitable cells, essential for the regulation of a plethora of Ca(2+)-dependent physio-pathological events, such as apoptosis in the presence of a Ca(2+) overload. There
Ana J Moreno-Ortega et al.
Neuropharmacology, 95, 503-510 (2015-04-25)
CALHM1 is a Ca(2+) channel discovered in 2008, which plays a key role in the neuronal electrical activity, among other functions. However, there are no known efficient blockers able to modulate its Ca(2+) handling ability. We herein describe that benzothiazepine
Francisco J Martínez-Sanz et al.
ACS chemical neuroscience, 6(9), 1626-1636 (2015-07-21)
Mitochondria regulate cellular Ca(2+) oscillations, taking up Ca(2+) through its uniporter and releasing it through the mitochondrial sodium/calcium exchanger. The role of mitochondria in the regulation of Ca(2+) cycle has received much attention recently, as it is a central stage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service