Skip to Content
Merck
All Photos(1)

Key Documents

SML1936

Sigma-Aldrich

Ro 64-6198

≥98% (HPLC)

Synonym(s):

(1S,3aS)-8-(2,3,3a,4,5,6-Hexahydro-1H-phenalen-1-yl)-1-phenyl-1,3,8-triazaspiro[4.5]decan-4-one, 8-[(1S,3aS)-2,3,3a,4,5,6-Hexahydro-1H-phenalen-1-yl]-1-phenyl-1,3,8-Triazaspiro[4.5]decan-4-one, Ro64-6198

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€149.00
25 MG
€599.00

€149.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€149.00
25 MG
€599.00

About This Item

Empirical Formula (Hill Notation):
C26H31N3O
CAS Number:
Molecular Weight:
401.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€149.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2(CCN([C@]3([H])C4=CC=CC5=C4[C@](CCC5)([H])CC3)CC2)N(C6=CC=CC=C6)CN1

InChI

1S/C26H31N3O/c30-25-26(29(18-27-25)21-9-2-1-3-10-21)14-16-28(17-15-26)23-13-12-20-7-4-6-19-8-5-11-22(23)24(19)20/h1-3,5,8-11,20,23H,4,6-7,12-18H2,(H,27,30)/t20-,23-/m0/s1

InChI key

JLFMYEAXZNPWBK-REWPJTCUSA-N

Biochem/physiol Actions

A brain-penetrant, nonpeptidic, high-affinity, reversible, selective nociceptin receptor ORL-1 (ORL1) agonist that elicits anxiolytic-like effects in vivo.
Ro 64-6198 is a brain-penetrant, nonpeptidic, high-affinity (Ki = 0.389 nM against 0.1 nM OFQ for binding human ORL1) nociceptin/orphanin FQ (N/OFQ) receptor ORL-1 (KOR-3, NOP) agonist (EC50 = 38.9 nM by GTPγS binding assay; IC50 = 32.4 nM against forskolin-stimulated cellular cAMP accumulation) with >100-fold selectivity over other opioid receptor family members (Ki = 46.8 nM/μ, 89.1 nM/k, 1.38 μM/δ) and no significant affinity toward 48 other receptors and ion channels (IC50 >1 μM). Ro 64-6198 elicits ORL1-dependent anxiolytic-like effects in several rat models of spontaneous and conditioned anxiety states (0.3-3.2 mg/kg i.p) without antipanic, anticonvulsant, sedative, amnestic activity, or signs of tolerance to its anxiolytic-like effects following chronical dosing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Jenck et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(9), 4938-4943 (2000-04-12)
The biochemical and behavioral effects of a nonpeptidic, selective, and brain-penetrant agonist at the ORL1 receptor are reported herein. This low molecular weight compound [(1S,3aS)-8- (2,3,3a,4,5, 6-hexahydro-1H-phenalen-1-yl)-1-phenyl-1,3,8-triaza- spiro[4. 5]decan-4-one] has high affinity for recombinant human ORL1 receptors and has 100-fold
F M Dautzenberg et al.
The Journal of pharmacology and experimental therapeutics, 298(2), 812-819 (2001-07-17)
The novel nonpeptide orphanin FQ/nociceptin (OFQ/N) ligand [(1S,3aS)-8-(2,3,3a,4,5,6-hexahydro-1H-phenalen-1-yl)-1-phenyl-1,3,8-triaza-spiro[4.5]decan-4-one] (Ro 64-6198) was characterized in vitro and in vivo for its agonistic potential. Ro 64-6198 was 130- to 3500-fold selective for the OFQ/N receptor (ORL1) compared with opiate receptors. In the cAMP
Phillip A Saccone et al.
The Journal of pharmacology and experimental therapeutics, 357(1), 17-23 (2016-01-24)
Nociceptin/orphanin FQ receptor (NOP) agonists have been reported to produce antinociceptive effects in rhesus monkeys with comparable efficacy to μ-opioid receptor (MOP) agonists, but without their limiting side effects. There are also known to be species differences between rodents and
Zhenlong Li et al.
The international journal of neuropsychopharmacology, 26(7), 483-495 (2023-06-07)
BTRX-246040, a nociceptin/orphanin FQ peptide receptor antagonist, is being developed for the treatment of depressive patients. However, the underlying mechanism of this potential antidepressant is still largely unclear. Here, we studied the antidepressant-related actions of BTRX-246040 in the ventrolateral periaqueductal
Celia Goeldner et al.
Psychopharmacology, 222(2), 203-214 (2012-01-18)
Ro 64-6198, the prototypical non-peptide nociceptin/orphanin FQ peptide (NOP) receptor agonist, has potent anxiolytic-like effects in several preclinical models and species. However the effects of Ro 64-6198 on distinctive anxiety-provoking conditions related to unconditioned conflict behavior as well as its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service