Skip to Content
Merck
All Photos(1)

Key Documents

SML1910

Sigma-Aldrich

IC87201

≥98% (HPLC)

Synonym(s):

2-[(1H-Benzotriazol-6-ylamino)methyl]-4,6-dichloro-phenol, 2-[(1H-Benzotriazol-6-ylamino)methyl]-4,6-dichlorophenol, IC 87201, IC-87201, Phenol, 2-[(1H-benzotriazol-6-ylamino)methyl]-4,6-dichloro-

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€67.40

€67.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€67.40

About This Item

Empirical Formula (Hill Notation):
C13H10Cl2N4O
CAS Number:
Molecular Weight:
309.15
UNSPSC Code:
12352200
NACRES:
NA.77

€67.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(c(cc(c1)Cl)CNc2cc3[nH]nnc3cc2)O

InChI

1S/C13H10Cl2N4O/c14-8-3-7(13(20)10(15)4-8)6-16-9-1-2-11-12(5-9)18-19-17-11/h1-5,16,20H,6H2,(H,17,18,19)

InChI key

QEHVTUCLCBXQIC-UHFFFAOYSA-N

Biochem/physiol Actions

IC87201 is a potent inhibitor of the nNOS/PSD-95 protein-protein interaction that blocks NMDA-induced 3′ ,5′ -cyclic guanosine monophosphate (cGMP) production in hippocampal cultures.
IC87201 is a potent inhibitor of the nNOS/PSD-95 protein-protein interaction that blocks NMDA-induced 3′ ,5′ -cyclic guanosine monophosphate (cGMP) production in hippocampal cultures. IC87201 exhibits analgesic properties in mice pain models. Contrary to NMDA receptor antagonists nNOS/PSD-95 protein-protein interaction inhibitors spared source memory, spatial memory, and motor function in rats.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra E Smith et al.
Behavioural brain research, 305, 23-29 (2016-02-26)
Limitations of preclinical models of human memory contribute to the pervasive view that rodent models do not adequately predict therapeutic efficacy in producing cognitive impairments or improvements in humans. We used a source-memory model (i.e., a representation of the origin
S K Florio et al.
British journal of pharmacology, 158(2), 494-506 (2009-09-08)
Post-synaptic density protein 95 (PSD95) contains three PSD95/Dosophilia disc large/ZO-1 homology domains and links neuronal nitric oxide synthase (nNOS) with the N-methyl-D-aspartic acid (NMDA) receptor. This report assesses the effects of disruption of the protein-protein interaction between nNOS and PSD95
Anders Bach et al.
Scientific reports, 5, 12157-12157 (2015-07-17)
ZL006 and IC87201 have been presented as efficient inhibitors of the nNOS/PSD-95 protein-protein interaction and shown great promise in cellular experiments and animal models of ischemic stroke and pain. Here, we investigate the proposed mechanism of action of ZL006 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service