Skip to Content
Merck
All Photos(1)

Documents

SML1790

Sigma-Aldrich

Tariquidar

≥98% (HPLC)

Synonym(s):

N-[2-[[[4-[2-(3,4-Dihydro-6,7-dimethoxy-2(1H)-isoquinolinyl)ethyl]phenyl]amino]carbonyl]-4,5-dimethoxyphenyl]-3-quinolinecarboxamide, XR 9576, XR9576

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H38N4O6
CAS Number:
Molecular Weight:
646.73
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC(OC)=C(OC)C=C1NC(C2=CC(C=CC=C3)=C3N=C2)=O)NC(C=C4)=CC=C4CCN5CC6=CC(OC)=C(OC)C=C6CC5

InChI

1S/C38H38N4O6/c1-45-33-18-25-14-16-42(23-28(25)19-34(33)46-2)15-13-24-9-11-29(12-10-24)40-38(44)30-20-35(47-3)36(48-4)21-32(30)41-37(43)27-17-26-7-5-6-8-31(26)39-22-27/h5-12,17-22H,13-16,23H2,1-4H3,(H,40,44)(H,41,43)

InChI key

LGGHDPFKSSRQNS-UHFFFAOYSA-N

Biochem/physiol Actions

Specific inhibitor of MDR-1 (P-gp)
Tariquidar is a non-competitive inhibitor. It is used as a potential agent for checking the role of P-glycoprotein inhibition in cancer. Tariquidar also has an ability to enhance drug exposure in drug-resistant cancers.
Tariquidar is a potent MDR1 (P-gp, P-glycoprotein) inhibitor. Tariquidar reverses multidrug resistance (MDR) by inhibiting MDR1-mediated drug efflux.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuemin Celina Chee et al.
Developmental cell, 46(6), 681-695 (2018-08-28)
The gut absorbs dietary nutrients and provides a barrier to xenobiotics and microbiome metabolites. To cope with toxin exposures, the intestinal epithelium is one of the most rapidly proliferating tissues in the body. The stem cell niche supplies essential signaling factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service