Skip to Content
Merck
All Photos(1)

Key Documents

SML1709

Sigma-Aldrich

K6PC-5

≥98% (HPLC)

Synonym(s):

2-Hexyl-N-[2-hydroxy-1-(hydroxymethyl)ethyl]-3-oxo-decanamide, K 6PC5;, N-(1,3-Dihydroxypropyl)-2-hexyl-3-oxo-decanamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€129.00
25 MG
€521.00

€129.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€129.00
25 MG
€521.00

About This Item

Empirical Formula (Hill Notation):
C19H37NO4
CAS Number:
Molecular Weight:
343.50
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€129.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(CCCCCCC)C(CCCCCC)C(NC(CO)CO)=O

InChI key

CGYVFCHCRBGGJG-UHFFFAOYSA-N

Biochem/physiol Actions

K6PC-5 is a potent and selective sphingosine kinase 1 (SphK1 or SK1) activator that increases keratin 1 and involucrin expression in normal human epidermal keratinocytes cultured in vitro. K6PC-5 promotes differentiation and proliferation of keratinocytes via intracellular Ca2+ signaling.
K6PC-5 is a potent and selective sphingosine kinase 1 (SphK1 or SK1) activator.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela T Reichmann et al.
The Journal of clinical investigation, 131(15) (2021-06-16)
Tuberculosis (TB) is a persistent global pandemic, and standard treatment for it has not changed for 30 years. Mycobacterium tuberculosis (Mtb) has undergone prolonged coevolution with humans, and patients can control Mtb even after extensive infection, demonstrating the fine balance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service