Skip to Content
Merck
All Photos(2)

Key Documents

857393P

Avanti

THI

2-acetyl-5-tetrahydroxybutyl imidazole, powder

Synonym(s):

2-acetyl-4-tetrahydroxybutyl imidazole; 2-acetyl-4(5)-tetrahydroxybutyl imidazole; 2-ATHBI

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€199.00

€199.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€199.00

About This Item

Empirical Formula (Hill Notation):
C9H14N2O5
CAS Number:
Molecular Weight:
230.22
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€199.00


Estimated to ship on18 May 2025


Request a Bulk Order

Assay

>98% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (857393P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857393P

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[nH]1c(ncc1[C@@H](O)[C@H](O)[C@H](O)CO)C(=O)C

InChI

1S/C9H14N2O5/c1-4(13)9-10-2-5(11-9)7(15)8(16)6(14)3-12/h2,6-8,12,14-16H,3H2,1H3,(H,10,11)/t6-,7-,8-/m1/s1

InChI key

CQSIXFHVGKMLGQ-BWZBUEFSSA-N

General description

2-Acetyl-5-tetrahydroxybutyl imidazole (THI) is an inhibitor of sphingosine-1-phosphate lyase (S1P lyase).[1]
Sphingosine-1-phosphate (S1P) lyase catalyzes the irreversible decomposition of S1P to hexadecanaldehyde and phosphoethanolamine. 2-Acetyl-5-tetrahydroxybutyl imidazole (THI) is an inhibitor of S1P lyase.[2] Because S1P lyase is primarily expressed in lymphatic tissue, treatment of mice with THI (50 μg/ml in drinking water) increases lymphoid tissue S1P concentrations 100-fold, reducing lymphocyte egress from thymus and peripheral lymphoid organs.[2][3] The resulting lymphopenia is reversible following cessation of THI treatment.[4] Reducing S1P lyase activity results in therapeutic levels of immunosuppression without the non-lymphoid lesions that result from synthetic S1P receptor agonists.[5]

Biochem/physiol Actions

2-Acetyl-5-tetrahydroxybutyl imidazole (THI) can block S1P lyase in vivo and up-regulates circulatory and tissue S1P levels.[6] It has the ability to repress dystrophic muscle degeneration.[7]

Packaging

5 mL Amber Glass Screw Cap Vial (857393P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The immunosuppressive compound 2-acetyl-4-tetrahydroxybutyl imidazole inhibits the allogeneic mixed lymphocyte reaction by sequestration of a recirculating subpopulation of T cells.
Bradbury MG, et al.
Immunology, 87, 80-85 (1996)
2-Acetyl-5-tetrahydroxybutyl imidazole (THI) protects 661W cells against oxidative stress
Fabiani C, et al.
Naunyn-Schmiedeberg'S Archives of Pharmacology, 390(7), 741-751 (2017)
Emigration of mature T cells from the thymus is inhibited by the imidazole-based compound 2-acetyl-4-tetrahydroxybutylimidazole.
Gugasyan R, et al.
Immunology, 93, 398-404 (1998)
Molecular mechanism of sphingosine-1-phosphate action in Duchenne muscular dystrophy
Nguyen TDH, et al.
Disease models & mechanisms, 7(1), 41-54 (2014)
Incomplete inhibition of sphingosine 1-phosphate lyase modulates immune system function yet prevents early lethality and non-lymphoid lesions.
Vogel P, et al.
PLoS ONE, 4(1) (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service