Skip to Content
Merck
All Photos(1)

Key Documents

SML1381

Sigma-Aldrich

PIK-294

≥98% (HPLC)

Synonym(s):

2-((4-Amino-3-(3-hydroxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)methyl)-5-methyl-3-o-tolylquinazolin-4(3H)-one, 2-[[4-Amino-3-(3-hydroxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]methyl]-5-methyl-3-(2-methylphenyl)-4(3H)-quinazolinone, PIK294

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€85.00
25 MG
€338.00

€85.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€85.00
25 MG
€338.00

About This Item

Empirical Formula (Hill Notation):
C28H23N7O2
CAS Number:
Molecular Weight:
489.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€85.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear
DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

NC1=C2C(N(CC(N3C4=C(C)C=CC=C4)=NC5=C(C(C)=CC=C5)C3=O)N=C2C6=CC(O)=CC=C6)=NC=N1

InChI

1S/C28H23N7O2/c1-16-7-3-4-12-21(16)35-22(32-20-11-5-8-17(2)23(20)28(35)37)14-34-27-24(26(29)30-15-31-27)25(33-34)18-9-6-10-19(36)13-18/h3-13,15,36H,14H2,1-2H3,(H2,29,30,31)

InChI key

WFSLJOPRIJSOJR-UHFFFAOYSA-N

Application

PIK-294 has been used as PI3K inhibitor in drug combination studies and their synergy quantification using the Chou-Talalay method.[1]

Biochem/physiol Actions

PIK-294 is a highly selective inihibitor of the phosphoinositide 3-kinase (PI3K) p110δ with an IC50 value of 10 nM compared to IC50 values of 10 μM, 490 nM and 160 nM for PI3Kα/β/γ, respectively. PIK-294 has been used to help distinguish the unique roles of the various PI3-K isoforms. For example, in a study of CXCL8-induced neutrophil migration, PIK-294 inhibited both chemokinetic and chemotactic CXCL8-induced migration whereas PI3Kγ inhibitor AS-605240 markedly reduced CXCL8 induced chemokinetic migration but had no effect on CXCL8 induced chemotactic migration.
PIK-294 is a highly selective inihibitor of the phosphoinositide 3-kinase (PI3K) p110δ.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raoni Pais Siqueira et al.
Toxicology in vitro : an international journal published in association with BIBRA, 65, 104777-104777 (2020-01-22)
The serine/arginine protein kinases respond to the EGFR-PI3K-AKT signaling module in the context of pre-mRNA alternative splicing regulation. These enzymes (notably SRPK1 and SRPK2) have been found dysregulated in a variety of cancers, which suggests them as promising drug targets

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service