Skip to Content
Merck
All Photos(1)

Key Documents

1.11956

Sigma-Aldrich

Hemacolor® Rapid staining of blood smear

Solution 2: colour reagent red

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
€146.00

€146.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
€146.00

About This Item

UNSPSC Code:
41116121
NACRES:
NA.21

€146.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

pH

6.8 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Hemacolor® Rapid staining of blood smear - Solution 2: colour reagent red, is a ready-to-use solution used for human-medical cell diagnosis and serves the fixing of native blood and bone-marrow smears of human origin in hematological and clinico-cytological specimen materials, for example smears of whole blood and bone marrow. The staining with Hemacolor® yields a staining result that corresponds to the Pappenheim stain, with predominantly magenta stained nuclei. This is based on the molecular interaction of the Eosin Y dye and a complex of Azur B with DNA. Both dyes assemble to an Eosin Y - Azur B-DNA complex and the intensity of the resulting stain depends on the content of Azur B and the ratio of Azur B : Eosin Y. Furthermore, the resulting stain can vary depending on the influence of fixation, staining times, pH-value of the solutions or buffer substances. By using pH 7.2 buffered solutions with the Hemacolor® Rapid staining of blood smear solutions, a high stability of the stain and clean, precipitation-free staining results can be guaranteed.
The package is sufficient for 1250 - 5000 applications. The buffer tablets acc. to Weise pH 7.2 (Cat no. 109468) are also available in larger pack sizes, thus, they can be useful for higher demands. The three solutions needed for staining are all available individually as:

Hemacolor® Rapid staining of blood smear solution 1, fixing solution (Product number 1.11955)
Hemacolor® Rapid staining of blood smear solution 2, colour reagent red (Product number 1.11956)
Hemacolor® Rapid staining of blood smear solution 3, colour reagent blue (Product number 1.11957)

or they can be purchased as staining sets:

Hemacolor® Rapid staining of blood smear (Product number 1.11674) 3 x 100 ml
Hemacolor® Rapid staining of blood smear (Product number 1.11661) 3 x 500 ml

This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Legal Information

HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can this kit also be used to produce blood smears where the blood was previously stored in EDTA tubes?

    1 answer
    1. Yes, the Hemacolor reagents are suitable for use with samples anticoagulated with EDTA. If the sample is not anticoagulated, the smears must be made immediately upon the blood being drawn. These reagents are not suitable for use with samples anticoagulated with heparin or sodium citrate. Smears should be prepared from fresh samples when prepared from anticoagulated blood with EDTA.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service