Skip to Content
Merck
All Photos(1)

Key Documents

SML1349

Sigma-Aldrich

SB-207266

≥98% (HPLC)

Synonym(s):

1′-Butyl-4′-piperidinylmethyl ester, 8-Amino-7-chloro-2,3-dihydro-1,4-benzodioxan-5-carboxylic acid, N-1-Butyl-4-piperinylmethyl)-3,4-dihydro-2H-[1,3] oxazino[3,2-a]indole-10-carboxamide, Piboserod

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€97.80
25 MG
€326.00

€97.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€97.80
25 MG
€326.00

About This Item

Empirical Formula (Hill Notation):
C22H31N3O2
CAS Number:
Molecular Weight:
369.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€97.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(NCC1CCN(CCCC)CC1)C2=C3N(CCCO3)C4=C2C=CC=C4

InChI

1S/C22H31N3O2/c1-2-3-11-24-13-9-17(10-14-24)16-23-21(26)20-18-7-4-5-8-19(18)25-12-6-15-27-22(20)25/h4-5,7-8,17H,2-3,6,9-16H2,1H3,(H,23,26)

InChI key

KVCSJPATKXABRQ-UHFFFAOYSA-N

Biochem/physiol Actions

SB-207266 is a Serotonin 5-HT4 receptor antagonist in vivo.
SB-207266 is a potent, selective, orally active and long-acting 5-HT4 antagonist in vivo.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbora Waclawiková et al.
PLoS biology, 19(1), e3001070-e3001070 (2021-01-23)
Microbial conversion of dietary or drug substrates into small bioactive molecules represents a regulatory mechanism by which the gut microbiota alters intestinal physiology. Here, we show that a wide variety of gut bacteria can metabolize the dietary supplement and antidepressant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service