Skip to Content
Merck
All Photos(1)

Key Documents

5.06415

Sigma-Aldrich

SB-269970 Hydrochloride

Synonym(s):

SB-269970 Hydrochloride, 5-HT7 Serotonin receptor antagonist, SB-269970, SB 26,970, SB 26,970

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€205.00

€205.00


Estimated to ship on28 May 2025



Select a Size

Change View
10 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C18H28N2O3S · xHCl
CAS Number:
Molecular Weight:
352.49 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on28 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: soluble

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N2[C@H](CCC2)CCN3CCC(CC3)C)c1cc(ccc1)O.Cl

InChI

1S/C18H28N2O3S.ClH/c1-15-7-11-19(12-8-15)13-9-16-4-3-10-20(16)24(22,23)18-6-2-5-17(21)14-18;/h2,5-6,14-16,21H,3-4,7-13H2,1H3;1H/t16-;/m1./s1

InChI key

XQCJOYZLWFNDIO-PKLMIRHRSA-N

General description

A selective 5-HT7 receptor antagonist. Extremely potent in vivo and in vitro. Chronic or acute (even single dose) administration of SB269970 induces functional desensitization of the 5-HT7 receptor system, which precedes changes in the receptor density. This mechanism may be responsible for the rapid antidepressant-like effect of the 5-HT7 antagonist in animal models. SB-269970 is an analogue of SB-258719 and has been shown to have at least 100 fold selectivity versus all other 5-HT receptor subtypes except the human 5-HT5A receptor (pKi values are 8.9, 7.2 and 6.0 for 5-HT7A, 5-HT5A and 5-HT1B).

Biochem/physiol Actions

Primary Target
5-HT₇

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tokarski, K. et al. 2012. Pharmacol. Rep.64, 256.
Thomas. D. R. et al. 2000. Br. J. Pharmacol.130, 409.
Lovell, P. J. et al. 2000. J. Med. Chem.43, 342.

Forbes, I. T. et al. 1998. J. Med. Chem.41, 655.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service