Skip to Content
Merck
All Photos(1)

Key Documents

SML1337

Sigma-Aldrich

NQ301

≥98% (HPLC)

Synonym(s):

2-Chloro-3-(4-acetophenyl)-amino-1,4-naphthoquinone, 2-[(4-Acetylphenyl)amino]-3-chloro-1,4-naphthalenedione, Compound 211, NQ 301

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€99.10
25 MG
€341.00

€99.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€99.10
25 MG
€341.00

About This Item

Empirical Formula (Hill Notation):
C23H19N3O · HBr
CAS Number:
Molecular Weight:
434.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€99.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, faint red to dark red

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C(Cl)=C(NC2=CC=C(C(C)=O)C=C2)C(C3=CC=CC=C31)=O

InChI

1S/C18H12ClNO3/c1-10(21)11-6-8-12(9-7-11)20-16-15(19)17(22)13-4-2-3-5-14(13)18(16)23/h2-9,20H,1H3

InChI key

LSQZKIQSQHZVQS-UHFFFAOYSA-N

Biochem/physiol Actions

NQ301 is an antithrombotic agent. Recent study shows that NQ301 (Compound 211) is a highly potent, selective, non-competitive allosteric inhibitor of CD45 that selectively inhibits dephosphorylation of substrate Lck pY-505 versus Lck pY-393. NQ301 prevents T cell receptor-mediated activation of Lck, Zap-70 and MAPK, and IL-2 production in primary T-cells. NQ301 exhibits immunosuppressive activity in mice.
NQ301 is an antithrombotic agent; antiplatelet; highly potent, selective non-competitive allosteric inhibitor of CD45.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service