Skip to Content
Merck
All Photos(1)

Key Documents

SML3118

Sigma-Aldrich

Obatoclax mesylate

≥98% (HPLC)

Synonym(s):

2-(2-((3,5-Dimethyl-1H-pyrrol-2-yl)methylene)-3-methoxy-2H-pyrrol-5-yl)-1H-indole, methanesulfonate salt, GX15-070 mesylate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€78.50
50 MG
€253.00

€78.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€78.50
50 MG
€253.00

About This Item

Empirical Formula (Hill Notation):
C20H19N3O · CH4O3S
CAS Number:
Molecular Weight:
413.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€78.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

dark green to black

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=S(O)(C)=O.CC1=C(NC(C)=C1)/C=C2N=C(C3=CC4=C(N3)C=CC=C4)C=C\2OC

InChI

1S/C20H19N3O.CH4O3S/c1-12-8-13(2)21-16(12)10-19-20(24-3)11-18(23-19)17-9-14-6-4-5-7-15(14)22-17;1-5(2,3)4/h4-11,21-22H,1-3H3;1H3,(H,2,3,4)/b19-10-;

InChI key

ZFKXDVMHNXPEIY-PEZBNFGJSA-N

Biochem/physiol Actions

BH3 domain-targeting inhibitor against Bcl-2 family anti-apoptotic proteins with anti-cancer efficacy in vitro and in vivo.
Obatoclax (GX15–070) is a BH3 domain-targeting inhibitor against Bcl-2 family anti-apoptotic proteins, including A1/Bfl-1, Bcl-2, Bcl-B, Bcl-w, Bcl-XL, and MCL-1. Obatoclax induces cancer cell apoptosis in cultures (0.05-2 μM) and in vivo (1.25-10 mg/kg via im. ip. or iv. in mice) by overcoming Bcl-2 members-mediated resistance to BAX- or BAK-dependent apoptosis.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service