Skip to Content
Merck
All Photos(1)

Key Documents

SML1274

Sigma-Aldrich

RI-1

≥98% (HPLC)

Synonym(s):

3-Chloro-1-(3,4-dichlorophenyl)-4-(4-morpholinyl)-1H-pyrrole-2,5-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€105.00
25 MG
€426.00

€105.00


Check Cart for Availability


Select a Size

Change View
5 MG
€105.00
25 MG
€426.00

About This Item

Empirical Formula (Hill Notation):
C14H11Cl3N2O3
CAS Number:
Molecular Weight:
361.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€105.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C=CC(N2C(C(N3CCOCC3)=C(Cl)C2=O)=O)=C1

InChI

1S/C14H11Cl3N2O3/c15-9-2-1-8(7-10(9)16)19-13(20)11(17)12(14(19)21)18-3-5-22-6-4-18/h1-2,7H,3-6H2

InChI key

MWSUIZKGNWELRF-UHFFFAOYSA-N

Biochem/physiol Actions

RI-1 is a cell-permeable RAD51 inhibitor that binds covalently to the protein surface at Cysteine 319. RI-1 disrupts homologous recombination in human cells.
RI-1 is a cell-permeable RAD51 inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd W Lewis et al.
Nucleic acids research, 47(7), 3503-3520 (2019-02-05)
The primary function of the UBE2T ubiquitin conjugase is in the monoubiquitination of the FANCI-FANCD2 heterodimer, a central step in the Fanconi anemia (FA) pathway. Genetic inactivation of UBE2T is responsible for the phenotypes of FANCT patients; however, a FANCT

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service