Skip to Content
Merck
All Photos(1)

Key Documents

SML0154

Sigma-Aldrich

SPV-106

≥98% (HPLC)

Synonym(s):

2-Pentadecylidene-Propanedioic acid 1,3-diethyl ester, Pentadecylidenemalonate 1b

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€125.00
25 MG
€487.00

€125.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€125.00
25 MG
€487.00

About This Item

Empirical Formula (Hill Notation):
C22H40O4
CAS Number:
Molecular Weight:
368.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€125.00


Estimated to ship on19 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCC\C=C(\C(=O)OCC)C(=O)OCC

InChI

1S/C22H40O4/c1-4-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21(23)25-5-2)22(24)26-6-3/h19H,4-18H2,1-3H3

InChI key

APIJHPAXMCKJBY-UHFFFAOYSA-N

Biochem/physiol Actions

Mixed histone acetylase inhibitor/activator
SPV106 is a mixed inhibitor/activator of histone acetyltransferase (HAT) activity. The compound inhibits the activity of p300/CBP, but potentiates p300/CBP-associated factor (PCAF) activity. SPV106 also induces S-phase cell cycle arrest and apoptosis in U937 cells.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service