Skip to Content
Merck
All Photos(1)

Key Documents

SML1166

Sigma-Aldrich

Salirasib

≥98% (HPLC)

Synonym(s):

2-(((2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrienyl)sulfanyl)benzoic acid, FTS, Farnesylthiosalicylic acid, S-Farnesylthiosalicylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€81.90
50 MG
€315.00

€81.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€81.90
50 MG
€315.00

About This Item

Empirical Formula (Hill Notation):
C22H30O2S
CAS Number:
Molecular Weight:
358.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€81.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(O)C1=CC=CC=C1SC/C=C(C)/CC/C=C(C)/CCC=C(C)C

InChI

1S/C22H30O2S/c1-17(2)9-7-10-18(3)11-8-12-19(4)15-16-25-21-14-6-5-13-20(21)22(23)24/h5-6,9,11,13-15H,7-8,10,12,16H2,1-4H3,(H,23,24)/b18-11+,19-15+

InChI key

WUILNKCFCLNXOK-CFBAGHHKSA-N

Application

Salirasib has been used as a farnesyltransferase inhibitor.[1]

Biochem/physiol Actions

Salirasib (Farnesylthiosalicylic acid) is a RAS inhibitor that acts by dislodging the farnesylated protein from the membrane, facilitating Ras degradation. Salirasib impairs downstream signaling and suppresses growth and migration of proliferating tumor cells in in vitro and in vivo models. Salirasib (Farnesylthiosalicylic acid) has recently been shown to possess significant anti-inflammatory and anti-arthritic properties.
Salirasib (Farnesylthiosalicylic acid) is a RAS inhibitor.
Salirasib (S-trans,trans-Farnesylthiosalicylic Acid /FTS) is a synthetic small molecule that has an ability to block all forms of Ras, unlike farnesyltransferase inhibitors, which fail to inhibit K-Ras and N-Ras function due to alternative membrane-binding mechanisms.[2][3] Salirasib along with gemcitabine, exhibits anti-tumor activity and biomarker modulation in preclinical models of metastatic pancreatic adenocarcinoma (PDA).[4] It is also used as a potent therapeutic for lung cancer.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane localization of acetylated CNK1 mediates a positive feedback on RAF/ERK signaling.
Fischer A, et al.
Science Advances, 3(8), e1700475-e1700475 (2017)
Orally administered FTS (salirasib) inhibits human pancreatic tumor growth in nude mice.
Haklai R, et al.
Cancer Chemotherapy and Pharmacology, 61(1), 89-96 (2008)
Suppression of lung cancer tumor growth in a nude mouse model by the Ras inhibitor salirasib (farnesylthiosalicylic acid).
Zundelevich A, et al.
Molecular Cancer Therapeutics, 6(6), 1765-1773 (2007)
Corina Beerli et al.
Nature microbiology, 4(2), 216-225 (2018-11-14)
Cell motility is essential for viral dissemination1. Vaccinia virus (VACV), a close relative of smallpox virus, is thought to exploit cell motility as a means to enhance the spread of infection1. A single viral protein, F11L, contributes to this by
A phase II trial of Salirasib in patients with lung adenocarcinomas with KRAS mutations.
Riely G J, et al.
Journal of Thoracic Oncology : Official Publication of the International Association for the Study of Lung Cancer, 6(8), 1435-1437 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service