Skip to Content
Merck
All Photos(1)

Documents

SML1163

Sigma-Aldrich

UNC2881

≥98% (HPLC)

Synonym(s):

2-(Butylamino)-4-[(trans-4-hydroxycyclohexyl)amino]-N-[[4-(1H-imidazol-1-yl)phenyl]methyl]-5-pyrimidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H33N7O2
CAS Number:
Molecular Weight:
463.58
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@@H](CC1)CC[C@H]1NC2=C(C(NCC3=CC=C(N4C=NC=C4)C=C3)=O)C=NC(NCCCC)=N2

InChI

1S/C25H33N7O2/c1-2-3-12-27-25-29-16-22(23(31-25)30-19-6-10-21(33)11-7-19)24(34)28-15-18-4-8-20(9-5-18)32-14-13-26-17-32/h4-5,8-9,13-14,16-17,19,21,33H,2-3,6-7,10-12,15H2,1H3,(H,28,34)(H2,27,29,30,31)/t19-,21-

InChI key

NPVXOWLPOFYACO-XUTJKUGGSA-N

Biochem/physiol Actions

UNC2881 is a selective inhbitor of Mer kinase, a member of the TAM (Tyro3, Axl, Mer) receptor tyrosine kinase family. The IC50 values for inhibition of Mer, Axl and Tyro 3 are 4.3, 360 and 250 nM, respectively.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service