Skip to Content
Merck
All Photos(1)

Key Documents

SML1158

Sigma-Aldrich

(2S)-Hypusine dihydrochloride

≥95% (HPLC)

Synonym(s):

(+)-Hypusine dihydrochloride, N6-[(2RS)-4-Amino-2-hydroxybutyl]-L-lysine dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€193.00
25 MG
€520.00

€193.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€193.00
25 MG
€520.00

About This Item

Empirical Formula (Hill Notation):
C10H23N3O3 · 2HCl
Molecular Weight:
306.23
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

€193.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

[H]Cl.[H]Cl.N[C@H](C(O)=O)CCCCNCC(O)CCN

InChI

1S/C10H23N3O3.2ClH/c11-5-4-8(14)7-13-6-2-1-3-9(12)10(15)16;;/h8-9,13-14H,1-7,11-12H2,(H,15,16);2*1H/t8?,9-;;/m0../s1

InChI key

UTSZZUVUESAPQD-AHBBCRTASA-N

Biochem/physiol Actions

(2S)-Hypusine is an unusual amino acid found only in eukaryotic translation initiation factor 5A (eIF5A) and a similar protein in archaebacteria. (2S)-Hypusine is formed by posttranslational modification of a lysine with the addition of the 4-aminobutyl moiety from the polyamine spermidine. There is increasing interest in the polyamine-hypusine axis and eIF5A, its a role in cell growth and differentiation, and a recent paper on the polyamine-hypusine axis as a potential new tumour suppressor network.
(2S)-Hypusine is an unusual amino acid found only in eukaryotic translation initiation factor 5A (eIF5A).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service