Skip to Content
Merck
All Photos(1)

Key Documents

SML1086

Sigma-Aldrich

BX-912

≥95% (HPLC)

Synonym(s):

N-[3-[[5-bromo-4-[[2-(1H-imidazol-5-yl)ethyl]amino]-2-pyrimidinyl]amino]phenyl]-1-pyrrolidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23BrN8O
CAS Number:
Molecular Weight:
471.35
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

Brc1c(nc(nc1)Nc3cc(ccc3)NC(=O)N4CCCC4)NCCc2nc[nH]c2

InChI key

DMMILYKXNCVKOJ-UHFFFAOYSA-N

Biochem/physiol Actions

BX-912 is a potent inhibitor of PDK1 (3-Phosphoinositide dependent protein kinase-1). The IC50 values for direct inhibition of kinase activity or inhibition in a cell based assay are 26 nM and 12 nM, respectively. BX-912 inhibits the growth of PC-3 tumor cells grown on soft agar more potently than cells grown on plastic (IC50 = 320 nM vs. 5.5 mM, respectively).
PDK1 inhibitor

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Munir A Al-Zeer et al.
EBioMedicine, 23, 100-110 (2017-08-15)
The intracellular human bacterial pathogen Chlamydia trachomatis pursues effective strategies to protect infected cells against death-inducing stimuli. Here, we show that Chlamydia trachomatis infection evokes 3-phosphoinositide-dependent protein kinase-1 (PDPK1) signaling to ensure the completion of its developmental cycle, further leading
Sameer S Chopra et al.
Cell systems, 10(1), 66-81 (2019-12-10)
Frequent mutation of PI3K/AKT/mTOR signaling pathway genes in human cancers has stimulated large investments in targeted drugs but clinical successes are rare. As a result, many cancers with high PI3K pathway activity, such as triple-negative breast cancer (TNBC), are treated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service