Skip to Content
Merck

Skip To

SML0987

CCG-1423

≥98% (HPLC)

Synonym(s):

CCG 1423, N-[2-[(4-Chlorophenyl)amino]-1-methyl-2-oxoethoxy]-3,5-bis(trifluoromethyl)-benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 132,00

About This Item

Empirical Formula (Hill Notation):
C18H13ClF6N2O3
CAS Number:
Molecular Weight:
454.75
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

€ 132,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(cc(c1)C(=O)NOC(C)C(=O)Nc2ccc(cc2)Cl)C(F)(F)F

InChI

1S/C18H13ClF6N2O3/c1-9(15(28)26-14-4-2-13(19)3-5-14)30-27-16(29)10-6-11(17(20,21)22)8-12(7-10)18(23,24)25/h2-9H,1H3,(H,26,28)(H,27,29)

InChI key

DSMXVSGJIDFLKP-UHFFFAOYSA-N

Biochem/physiol Actions

CCG-1423 is a potent and specific inhibitor of Rho pathway signaling and activation of serum response factor (SRF) transcription.
CCG-1423 is a potent and specific inhibitor of Rho pathway signaling and activation of serum response factor (SRF) transcription. CCG-1423 inhibits SRF-driven luciferase expression in PC-3 cells following stimulation with constitutively active Gα13. Also CCG-1423 selectively inhibits DNA synthesis, proliferation and invasion of Rho-overexpressing cell lines.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
555558G7423SML2411
description

≥98% (HPLC)

description

The Rho/SRF Pathway Inhibitor, CCG-1423, also referenced under CAS 285986-88-1, controls the biological activity of Rho/SRF. This small molecule/inhibitor is primarily used for Cell Structure applications.

description

≥98% (HPLC), white to off-white, powder

description

≥98% (HPLC)

form

powder

form

solid

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 10 mg/mL

solubility

DMSO: ≥10 mg/mL

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white

color

white to off-white

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters