Skip to Content
Merck
All Photos(1)

Key Documents

SML2411

Sigma-Aldrich

KPR-2579

≥98% (HPLC)

Synonym(s):

(αR)-α-[Benzoyl[(1R)-1-(3,5-difluorophenyl)ethyl]amino]-benzeneacetamide, N-((R)-2-Amino-2-oxo-1-phenylethyl)-N-((R)-1-(3,5-difluorophenyl)ethyl)benzamide, N-((R)-Carbamoylphenylmethyl)-N-[(R)-1-(3,5-difluorophenyl)ethyl]benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€55.80
25 MG
€225.00

€55.80


Estimated to ship on11 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€55.80
25 MG
€225.00

About This Item

Empirical Formula (Hill Notation):
C23H20F2N2O2
CAS Number:
Molecular Weight:
394.41
UNSPSC Code:
12352200

€55.80


Estimated to ship on11 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +124 to +136°, c = c=1 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C1)N([C@H](C)C2=CC(F)=CC(F)=C2)[C@H](C3=CC=CC=C3)C(N)=O

InChI

1S/C23H20F2N2O2/c1-15(18-12-19(24)14-20(25)13-18)27(23(29)17-10-6-3-7-11-17)21(22(26)28)16-8-4-2-5-9-16/h2-15,21H,1H3,(H2,26,28)/t15-,21-/m1/s1

InChI key

NNXPFEMWLZLAHM-QVKFZJNVSA-N

Biochem/physiol Actions

KPR-2579 does not distress temperature of the body at any given dose. It helps to block the activation of C-fiber single-unit afferent activities (SAAs), stimulated by acetic acid (AA).[1]
KPR-2579 is a potent and selective transient receptor potential melastatin 8 (TRPM8; CRM1) antagonist (IC50 = 80/89 nM against EC80 MeOH-induced Ca2+ response in human/rat CRM1 HEK293T transfectants) with good selectivity (IC50 >30 μM against ligand-induced Ca2+ influx using hTRPA1, hTRPV1, or hTRPV4 transfectants) and oral availability (59% post 10 mg/kg p.o.; rats). KPR-2579 exhibits in vivo efficacy against icilin-induced wet-dog shakes (WDS; by 31/73/100% with 1/3/10 mg/kg p.o. 1h prior to icilin i.p.; female rats) and distension-induced rhythmic bladder contraction (by 70/89% with 0.1/0.3 mg/kg i.v.; male rats) without negative cardiovascular effects.
Potent and selective transient receptor potential melastatin 8 (TRPM8; CRM1) antagonist with good selectivity, in vivo efficacy and oral bioavailability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Ichi Kobayashi et al.
Bioorganic & medicinal chemistry, 25(2), 727-742 (2016-12-15)
Transient receptor potential melastatin 8 (TRPM8) is activated by innocuous cold and chemical substances, and antagonists of this channel have been considered to be effective for pain and urinary diseases. N-(3-aminopropyl)-2-{[(3-methylphenyl)methyl]oxy}-N-(2-thienylmethyl)benzamide hydrochloride (AMTB), a TRPM8 antagonist, was proposed to be
KPR-2579, a novel TRPM8 antagonist, inhibits acetic acid-induced bladder afferent hyperactivity in rats
Aizawa N, et al.
Neurourology and Urodynamics, 37(5), 1633-1640 (2018)
Naoki Aizawa et al.
Neurourology and urodynamics, 37(5), 1633-1640 (2018-02-22)
Transient receptor potential melastatin 8 (TRPM8) is proposed to be a promising therapeutic target for hypersensitive bladder disorders. We examined the effects of KPR-2579, a novel selective TRPM8 antagonist, on body temperature and on mechanosensitive bladder single-unit afferent activities (SAAs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service