Skip to Content
Merck
All Photos(1)

Key Documents

SML0979

Sigma-Aldrich

Bantag-1 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Boc-Phe-His-4-amino-5-cyclohexyl-2,4,5-trideoxypentonyl-Leu-(3-dimethylamino) benzylamide N-methylammonium trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€331.00

€331.00


Check Cart for Availability


Select a Size

Change View
1 MG
€331.00

About This Item

Empirical Formula (Hill Notation):
C48H73N8O7 · xC2HF3O2
Molecular Weight:
874.14 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€331.00


Check Cart for Availability

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

General description

Bantag-1 is a peptide-antagonist.[1] It is soluble in aqueous solutions.[2]

Biochem/physiol Actions

Bantag-1 is a very specific, high affinity antagonist of the orphan GPCR Bombesin receptor subtype-3 (BRS-3).
Bantag-1 is a very specific, high affinity antagonist of the orphan GPCR Bombesin receptor subtype-3 (BRS-3). The binding affinity of Bantag-1 in BRS-3 expressing cells is 1.3 nM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular basis for high affinity and selectivity of peptide antagonist, Bantag-1, for the orphan BB3 receptor.
Nakamura T, et al.
Biochemical Pharmacology, 115, 64-76 (2016)
Xiao-Ming Guan et al.
Cell metabolism, 11(2), 101-112 (2010-01-26)
Bombesin receptor subtype 3 (BRS-3) is a G protein coupled receptor whose natural ligand is unknown. We developed potent, selective agonist (Bag-1, Bag-2) and antagonist (Bantag-1) ligands to explore BRS-3 function. BRS-3-binding sites were identified in the hypothalamus, caudal brainstem

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service