Skip to Content
Merck
All Photos(1)

Key Documents

SML1539

Sigma-Aldrich

TDFA trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

N-Acetyl-L-threonyl-L-α-aspartyl-N5-(2-fluoro-1-iminoethyl)-L-ornithinamide trifluoroacetate salt, Thr-Asp-F-amidine trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€297.00
25 MG
€1,200.00

€297.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€297.00
25 MG
€1,200.00

About This Item

Empirical Formula (Hill Notation):
C17H29FN6O7 · xC2HF3O2
Molecular Weight:
448.45 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€297.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL, clear

shipped in

dry ice

storage temp.

−70°C

General description

TDFA trifluoroacetate salt or tripeptide F-amidine analog is a derivative synthesized to increase the selectivity, bioavailability and potency of Cl-amidine, which is a member of protein arginine deiminases (PADs) compounds.[1]

Biochem/physiol Actions

Selective inhibitor of PAD4 (peptidyl arginine deiminase 4)
TDFA (Thr-Asp-F-amidine) is a selective inhibitor of peptidyl arginine deiminase 4 (protein arginine deiminase 4, PAD4 or PADI4), a transcriptional coregulator that catalyzes the calcium-dependent conversion of specific arginine residues in proteins to citrulline, a process called citrullination. PAD4 is upregulated in cancer and inflammatory diseases, and may be involved in rheumatoid arthritis onset and progression. It has recently been found to be involved in the regulation of pluripotency: important for stem-cell gene activation, induced pluripotent stem (iPS) cell reprogramming, and maintenance of pluripotent cells in the early mouse embryo. TDFA has an IC50 value of 2.3 μM and is ≥15-fold selective for PAD4 versus PAD1 and ≥50-fold selective versus PADs 2 and 3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis, and biological evaluation of tetrazole analogs of Cl-amidine as protein arginine deiminase inhibitors.
Subramanian V
Journal of Medicinal Chemistry, 58(3), 1337-1344 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service