Skip to Content
Merck

Skip To

SML0915

Xipamide

≥98% (HPLC)

Synonym(s):

5-(Aminosulfonyl)-4-chloro-N-(2,6-dimethylphenyl)-2-hydroxy-benzamide, BE 1293, BEI 1293, MJF 10938

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 212,00

About This Item

Empirical Formula (Hill Notation):
C15H15ClN2O4S
CAS Number:
Molecular Weight:
354.81
UNSPSC Code:
12352200
EC Number:
238-216-4
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

€ 212,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(N)c1c(cc(c(c1)C(=O)Nc2c(cccc2C)C)O)Cl

InChI

1S/C15H15ClN2O4S/c1-8-4-3-5-9(2)14(8)18-15(20)10-6-13(23(17,21)22)11(16)7-12(10)19/h3-7,19H,1-2H3,(H,18,20)(H2,17,21,22)

InChI key

MTZBBNMLMNBNJL-UHFFFAOYSA-N

Biochem/physiol Actions

Xipamide is a sulfonamide diuretic that blocks sodium reabsorption in the distal tubules of the kidney, resulting in increased urine output. Xiopamide also blocks the cystic fibrosis transmembrane conductance regulator (CFTR) chloride channel.
Xipamide is a sulfonamide diuretic.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1604SML1066SML1616
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

storage condition

desiccated

storage condition

-

storage condition

-

storage condition

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters