Skip to Content
Merck
All Photos(1)

Key Documents

SML1616

Sigma-Aldrich

ICA-069673

≥98% (HPLC)

Synonym(s):

ICA 069673, N-(2-Chloro-5-pyrimidinyl)-3,4-difluoro-benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€104.00
25 MG
€417.00

€104.00


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
€104.00
25 MG
€417.00

About This Item

Empirical Formula (Hill Notation):
C11H6ClF2N3O
CAS Number:
Molecular Weight:
269.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€104.00


Estimated to ship on31 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(C1=CC=C(F)C(F)=C1)NC2=CN=C(Cl)N=C2

InChI

1S/C11H6ClF2N3O/c12-11-15-4-7(5-16-11)17-10(18)6-1-2-8(13)9(14)3-6/h1-5H,(H,17,18)

InChI key

IIBSHMFXVWTQSJ-UHFFFAOYSA-N

Biochem/physiol Actions

ICA-069673 has higher selectivity towards KV7.2/KV7.3 heterodimeric channels than KV7.3/KV7.5 channels. ICA-069673 is considered as a part of gating modifiers.[1]
ICA-069673 is an orally active, potent and selective KV7.2/KV7.3 (KCNQ2/Q3) channel opener that is active in rodent epilepsy models.
ICA-069673 is an orally active, potent and selective KV7.2/KV7.3 (KCNQ2/Q3) channel opener.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The novel KV7. 2/KV7. 3 channel opener ICA-069673 reveals subtype-specific functional roles in guinea pig detrusor smooth muscle excitability and contractility.
Provence A, et al.
Journal of Pharmacology and Experimental Therapeutics, 354(3), 290-301 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service