Skip to Content
Merck
All Photos(1)

Key Documents

SML2328

Sigma-Aldrich

BD064

≥98% (HPLC)

Synonym(s):

B-[5-[[[1-[3-(4-Ethoxyphenyl)-3,4-dihydro-4-oxopyrido[2,3-d]pyrimidin-2-yl]ethyl][2-[4-fluoro-3-(trifluoromethyl)phenyl]acetyl]amino]methyl]-2-fluorophenyl]boronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€80.80
25 MG
€267.00

€80.80


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€80.80
25 MG
€267.00

About This Item

Empirical Formula (Hill Notation):
C33H28BF5N4O5
CAS Number:
Molecular Weight:
666.40
UNSPSC Code:
12352200
NACRES:
NA.77

€80.80


Estimated to ship on27 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1N(C2=CC=C(OCC)C=C2)C(C(N(C(CC3=CC=C(F)C(C(F)(F)F)=C3)=O)CC4=CC(B(O)O)=C(F)C=C4)C)=NC5=NC=CC=C51

Biochem/physiol Actions

BD064 is a probe-dependent and biased negative allosteric modulator (NAM) of the chemokine receptor CXCR3 signaling that preferentially inhibits CXCL11-mediated ?-arrestin 2 recruitment over G protein activation.
biased negative allosteric modulator of the chemokine receptor CXCR3 signaling that preferentially inhibits CXCL11-mediated β-arrestin 2 recruitment

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viachaslau Bernat et al.
ChemMedChem, 10(3), 566-574 (2015-02-07)
Over the last decade, functional selectivity (or ligand bias) has evolved from being a peculiar phenomenon to being recognized as an essential feature of synthetic ligands that target G protein-coupled receptors (GPCRs). The CXC chemokine receptor 3 (CXCR3) is an outstanding platform
Regine Brox et al.
Molecular pharmacology, 93(4), 309-322 (2018-01-19)
Our recent explorations of allosteric modulators with improved properties resulted in the identification of two biased negative allosteric modulators, BD103 (N-1-{[3-(4-ethoxyphenyl)-4-oxo-3,4-dihydropyrido[2,3-d]pyrimi-din2yl]ethyl}-4-(4-fluorobutoxy)-N-[(1-methylpiperidin-4-yl)methyl}]butanamide) and BD064 (5-[(N-{1-[3-(4-ethoxyphenyl)-4-oxo-3,4-dihydropyrido[2,3-d]pyrimidin-2-yl]ethyl-2-[4-fluoro-3-(trifluoromethyl)phenyl]acetamido)methyl]-2-fluorophenyl}boronic acid), that exhibited probe-dependent inhibition of CXC-motif chemokine receptor CXCR3 signaling. With the intention to elucidate
Viachaslau Bernat et al.
ACS chemical biology, 9(11), 2664-2677 (2014-09-19)
The chemokine receptor CXCR3 is a G protein-coupled receptor, which conveys extracellular signals into cells by changing its conformation upon agonist binding. To facilitate the mechanistic understanding of allosteric modulation of CXCR3, we combined computational modeling with the synthesis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service