Skip to Content
Merck
All Photos(1)

Key Documents

SML0898

Sigma-Aldrich

Mps1-IN-3

≥98% (HPLC)

Synonym(s):

1-(4-(6-(2-(Isopropylsulfonyl)phenylamino)-9H-purin-2-ylamino)-3-methoxyphenyl)piperidin-4-ol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€121.00

€121.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€121.00

About This Item

Empirical Formula (Hill Notation):
C26H31N7O4S
CAS Number:
Molecular Weight:
537.63
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€121.00


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1CCN(C2=CC=C(NC3=NC(NC=N4)=C4C(NC5=C(S(=O)(C(C)C)=O)C=CC=C5)=N3)C(OC)=C2)CC1

InChI

1S/C26H31N7O4S/c1-16(2)38(35,36)22-7-5-4-6-20(22)29-25-23-24(28-15-27-23)31-26(32-25)30-19-9-8-17(14-21(19)37-3)33-12-10-18(34)11-13-33/h4-9,14-16,18,34H,10-13H2,1-3H3,(H3,27,28,29,30,31,32)

InChI key

OFRMASLPWOMYHN-UHFFFAOYSA-N

General description

Monopolar spindle 1 (Mps1) protein contains a TPR (tetratricopeptide repeat) motif at its N-terminal and a C-terminal kinase domain.[1]

Application

Mps1-IN-3 has been used in 2D-proliferation and 3D-organotypic culture outgrowth assays.[2]

Biochem/physiol Actions

Mps1 mediates phosphorylation of kinetochore scaffold 1.[1]
Mps1-IN-3 is an inhibitor of Monopolar spindle 1 (MPS1, also known as TTK), a Dual-specificity kinase required for spindle pole body duplication and spindle checkpoint function. Mps1-IN-3 inhibited Mps1 kinase activity with an IC50 value of 50 nM inhibited the proliferation of U251 glioblastoma cells as determined by cell counting with an IC50 value of approximately 5 μM, and sensitized glioblastoma cells to vincristine in orthotopic mouse models.
Mps1-IN-3 is an inhibitor of Monopolar spindle 1 (MPS1.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Cancer Drug Targets: Volume: 2, 61-61 (2014)
Mutant p53 dictates the oncogenic activity of c-Abl in triple-negative breast cancers.
Morrison C D, et al.
Cell Death & Disease, 8(6), e2899-e2899 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service