Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200631

Sigma-Aldrich

Monoclonal Anti-CYBA antibody produced in mouse

clone CB-64, purified from hybridoma cell culture

Synonym(s):

Cytochrome b558 subunit alpha, Neutrophil cytochrome b 22 kDa polypeptide, Superoxide-generating NADPH oxidase light chain subunit, cytochrome b-245 alpha polypeptide, p22-PHOX

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€454.00

€454.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€454.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€454.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CB-64, monoclonal

form

buffered aqueous solution

species reactivity

bovine, mouse, rat, monkey, human, canine

concentration

~1 mg/mL

technique(s)

flow cytometry: 5-10 μg/test using HeLa cells.
immunoblotting: 0.5-1 μg/mL using whole extracts of HeLa cells.
immunofluorescence: 5-10 μg/mL using HeLa cells.

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYBA(1535)
mouse ... Cyba(13057)
rat ... Cyba(79129)

General description

Monoclonal Anti-CYBA (mouse IgM isotype) is derived from the hybridoma CB-64 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a sequence at the internal region of human CYBA, conjugated to keyhole limpet hemocyanin (KLH). The small subunit, p22phox subunit, also known as cytochrome b245, alpha polypeptide/light chain (CYBA) is membrane bound,which is expressed in phagocytic, endothelial and vascular smooth muscle cells. This gene is located on human chromosome16q24

Immunogen

Synthetic peptide corresponding to a sequence at the internal region of human CYBA .

Application

Monoclonal Anti-CYBA antibody produced in mouse has been used in:
  • immunoblotting
  • flow cytometry
  • immunofluorescence

Biochem/physiol Actions

In prostate cancer cells downregulation of cytochrome b245, alpha polypeptide/light chain (CYBA) was reported to inhibited cell proliferation and colony formation, through AKT and ERK1/2 signaling pathways, indicating CYBA has a role in tumor angiogenesis and tumor growth. Mutations in this gene causes an autosomal recessive forms of chronic granulomatous disease (AR220CGD).

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NADPH oxidase subunit p22phox-mediated reactive oxygen species contribute to angiogenesis and tumor growth through AKT and ERK1/2 signaling pathways in prostate cancer
Li Q, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1833(12) (2013)
Three novel mutations in CYBA among 22 Iranians with Chronic granulomatous disease
Badalzadeh M, et al.
International Journal of Immunogenetics, 44(6) (2017)
CYBA encoding p22phox, the cytochrome b558 alpha polypeptide: gene structure, expression, role and physiopathology
Stasia MJ
Gene, 586(1) (2016)
Is the C242T Polymorphism of the CYBA Gene Linked with Oxidative Stress-Associated Complications of Prematurity
Huizing MJ, et al.
Antioxidants & Redox Signaling, 27(17) (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service