Skip to Content
Merck
All Photos(1)

Key Documents

SML0836

Sigma-Aldrich

ML297

≥98% (HPLC)

Synonym(s):

1-(3,4-Difluorophenyl)-3-(3-methyl-1-phenyl-1H-pyrazol-5-yl)urea, CID 56642816, VU0456810

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€476.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€476.00

About This Item

Empirical Formula (Hill Notation):
C17H14F2N4O
CAS Number:
Molecular Weight:
328.32
UNSPSC Code:
12352200
NACRES:
NA.77

€112.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

SMILES string

Fc1c(ccc(c1)NC(=O)Nc2[n](nc(c2)C)c3ccccc3)F

InChI

1S/C17H14F2N4O/c1-11-9-16(23(22-11)13-5-3-2-4-6-13)21-17(24)20-12-7-8-14(18)15(19)10-12/h2-10H,1H3,(H2,20,21,24)

InChI key

IEKSMUSSYJUQMY-UHFFFAOYSA-N

Biochem/physiol Actions

ML297 (VU0456810) is a potent and selective activator of the GIRK potassium channel.
ML297 (VU0456810) is the first potent and selective activator of the GIRK potassium channel. The G protein-coupled inwardly-rectifying potassium channels (GIRKs 1-4) are widely distributed broadly in the central nervous system (GIRKs 1-3) and also found in the heart (GIRK-4). ML297 activates GIRK channels containing a GIRK1 subunit, and does not activate channels without a GIRK1 subunit or the closely related potassium channel Kir2.1 or voltage-gated potassium channels. ML297 has been shown to activate GIRK1/2 with a potency of ~160 nM and also to activate GIRK channels comprising of GIRK1/3 and GIRK1/4 subunit combinations. ML297 was active in two in vivo models of epilepsy, and showed equal or greater efficacy than the anti-seizure medication, sodium valproate.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose Carlos Gonzalez et al.
Cell reports, 42(2), 112039-112039 (2023-02-08)
The central circadian regulator within the suprachiasmatic nucleus transmits time of day information by a diurnal spiking rhythm driven by molecular clock genes controlling membrane excitability. Most brain regions, including the hippocampus, harbor similar intrinsic circadian transcriptional machinery, but whether
Yulin Zhao et al.
Cell reports, 31(11), 107770-107770 (2020-06-20)
G-protein-gated inwardly rectifying K+ (GIRK) channels are essential effectors of inhibitory neurotransmission in the brain. GIRK channels have been implicated in diseases with abnormal neuronal excitability, including epilepsy and addiction. GIRK channels are tetramers composed of either the same subunit
Serap Erdogmus et al.
Nature communications, 10(1), 5784-5784 (2019-12-21)
G-protein coupled receptors (GPCRs) are versatile cellular sensors for chemical stimuli, but also serve as mechanosensors involved in various (patho)physiological settings like vascular regulation, cardiac hypertrophy and preeclampsia. However, the molecular mechanisms underlying mechanically induced GPCR activation have remained elusive.

Articles

We offer many products related to potassium channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service