Skip to Content
Merck

Skip To

5.08159

Salvinorin A

Synonym(s):

Salvinorin A, κ-opioid Receptor Agonist, Salvinorin A, Divinorin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 192,00

About This Item

Empirical Formula (Hill Notation):
C23H28O8
CAS Number:
Molecular Weight:
432.46
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

€ 192,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

Quality Segment

form

solid

potency

4.3 nM Ki

manufacturer/tradename

Calbiochem®

drug control

Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

storage condition

OK to freeze, protect from light

color

white

solubility

ethanol: 10 mM, DMSO: 50 mM

storage temp.

−70°C

SMILES string

[o]1cc(cc1)[C@H]2OC(=O)[C@H]3[C@@]([C@@H]4[C@]([C@@H](C[C@@H](C4=O)OC(=O)C)C(=O)OC)(CC3)C)(C2)C

InChI

1S/C23H28O8/c1-12(24)30-16-9-15(20(26)28-4)22(2)7-5-14-21(27)31-17(13-6-8-29-11-13)10-23(14,3)19(22)18(16)25/h6,8,11,14-17,19H,5,7,9-10H2,1-4H3/t14-,15-,16-,17-,19-,22-,23-/m0/s1

InChI key

OBSYBRPAKCASQB-AGQYDFLVSA-N

General description

A highly potent and selective non-alkaloid positive allosteric modulator of Κ-opioid receptors (Ki = 4.3 nM and 16 nM for native and cloned κ-opioid receptors, respectively). Also acts as a D2DR partial agonist (Ki = 5-10 nM). Used in studies of addition.

Biochem/physiol Actions

Primary Target
k opioid receptor

Other Notes

Roth, B. et al., 2002. PNAS.99, 11934.
Valdés III., j., et al., 1984. J. Organic Chem.49, 4716.
Ortega, A., et al., 1982. J. of Chem. Society, Perkins Transactions I. 2505.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.080175.081615.08316
form

solid

form

solid

form

solid

form

solid

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

drug control

Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

drug control

-

drug control

-

drug control

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under Salvinorin A


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters