Skip to Content
Merck
All Photos(1)

Key Documents

SML0794

Sigma-Aldrich

T2AA hydrochloride

≥98% (HPLC)

Synonym(s):

(S)-4-(4-(2-amino-3-hydroxypropyl)-2,6-diiodophenoxy)phenol hydrochloride, T2-aminoalcohol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€127.00
25 MG
€493.00

€127.00


Estimated to ship on29 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€127.00
25 MG
€493.00

About This Item

Empirical Formula (Hill Notation):
C15H15I2NO3 · HCl
CAS Number:
Molecular Weight:
547.55
UNSPSC Code:
12352200
NACRES:
NA.77

€127.00


Estimated to ship on29 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Ic1c(c(cc(c1)C[C@H](N)CO)I)Oc2ccc(cc2)O

InChI

1S/C15H15I2NO3/c16-13-6-9(5-10(18)8-19)7-14(17)15(13)21-12-3-1-11(20)2-4-12/h1-4,6-7,10,19-20H,5,8,18H2/t10-/m0/s1

InChI key

NKOSKXJRVWVXRI-JTQLQIEISA-N

Application

T2AA hydrochloride has been used as a chemical inhibitor of proliferating cell nuclear antigen (PCNA) in vaccinia virus.[1]

Biochem/physiol Actions

T2AA is an inhibitor of PCNA/ PIP-Box interactions that binds to the PCNA cavity that interacts with PIP-Box sequences. Also T2AA inhibits PCNA Polδ3 interaction. T2AA inhibits de novo DNA synthesis, and in combination of cisplatin significantly increases growth inhibition in cancer cells. T2AA is a T3 analog that exhibits almost no thyroid hormone activity.
T2AA is an inhibitor of PCNA/ PIP-Box interactions; inhibits PCNA Polδ3 interaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytoplasmic ATR activation promotes vaccinia virus genome replication.
Postigo A, et al.
Cell Reports, 19(5), 1022-1032 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service