Skip to Content
Merck
All Photos(1)

Key Documents

SML0666

Sigma-Aldrich

GSK1210151A

≥98% (HPLC)

Synonym(s):

7-(3,5-Dimethyl-4-isoxazolyl)-8-(methyloxy)-1-[(1R)-1-(2-pyridinyl)ethyl]-1,3-dihydro-2H-imidazo[4,5-c]quinolin-2-one, I-BET151

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€147.00
25 MG
€581.00

€147.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€147.00
25 MG
€581.00

About This Item

Empirical Formula (Hill Notation):
C23H21N5O3
CAS Number:
Molecular Weight:
415.44
UNSPSC Code:
12352200
NACRES:
NA.77

€147.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

originator

GlaxoSmithKline

relevant disease(s)

cancer

storage temp.

2-8°C

SMILES string

[n]2(c3c([nH][c]2=O)cnc4c3cc(c(c4)c5c(n[o]c5C)C)OC)[C@H](C)c1ncccc1

InChI

1S/C23H21N5O3/c1-12-21(14(3)31-27-12)16-9-18-15(10-20(16)30-4)22-19(11-25-18)26-23(29)28(22)13(2)17-7-5-6-8-24-17/h5-11,13H,1-4H3,(H,26,29)/t13-/m1/s1

InChI key

VUVUVNZRUGEAHB-CYBMUJFWSA-N

Application

GSK1210151A or I-BET151 has been used to study its inhibitory effect on BET (bromodomain and extra terminal) recruitment to chromatin in treating MLL (mixed-lineage leukemia)-fusion leukemia.[1]

Biochem/physiol Actions

GSK1210151A (I-BET151) is an inhibitor of the BET (bromodomain and extra terminal domain protein) family of acetyl-lysine recognizing, chromatin ′adaptor′ proteins.
GSK1210151A (I-BET151) is an inhibitor of the BET (bromodomain and extra terminal domain protein) family of acetyl-lysine recognizing, chromatin ′adaptor′ proteins. GSK1210151A displaces BRD3 and BRD4, PAFc and SEC components from chromatin resulting in inhibition of transcription at key genes (BCL2, C-MYC and CDK6) involved in the initiation of mixed lineage leukemia (MLL). GSK1210151A (I-BET151) showed good efficacy in 2 MLL animal models.

Other Notes

GSK1210151A has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the GSK1210151A probe summary on the Chemical Probes Portal website.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Replication Study: Inhibition of BET recruitment to chromatin as an effective treatment for MLL-fusion leukaemia.
Shan X, et al.
eLife, 6, e25306-e25306 (2017)
Lisa-Maria Winter et al.
Scientific reports, 13(1), 12061-12061 (2023-07-27)
GDF15 has recently emerged as a key driver of the development of various disease conditions including cancer cachexia. Not only the tumor itself but also adverse effects of chemotherapy have been reported to contribute to increased GDF15. Although regulation of
Zhongyuan Gao et al.
Cancer biology & therapy, 19(5), 407-415 (2018-01-16)
Lung cancer is the leading cause of cancer-related death worldwide. Bromodomain and extraterminal domain (BET) proteins act as epigenome readers for gene transcriptional regulation. Among BET family members, BRD4 was well studied, but for its mechanism in non-small cell lung
Yan-Yi Jiang et al.
Gastroenterology, 159(4), 1311-1327 (2020-07-04)
We investigated the transcriptome of esophageal squamous cell carcinoma (ESCC) cells, activity of gene regulatory (enhancer and promoter regions), and the effects of blocking epigenetic regulatory proteins. We performed chromatin immunoprecipitation sequencing with antibodies against H3K4me1, H3K4me3, and H3K27ac and
Jun Li et al.
Cell reports, 37(11), 110124-110124 (2021-12-16)
Regulatory T (Treg) cells play crucial roles in suppressing deleterious immune response. Here, we investigate how Treg cells are mechanistically induced in vitro (iTreg) and stabilized via transcriptional regulation of Treg lineage-specifying factor Foxp3. We find that acetylation of histone tails

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service