Skip to Content
Merck
All Photos(1)

Key Documents

SML0484

Sigma-Aldrich

KS-176

≥98% (HPLC)

Synonym(s):

N-[4-(2-Hydroxyethyl)phenyl]-2-[(4-nitrobenzoyl)amino]-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N3O5
CAS Number:
Molecular Weight:
405.40
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

OCCc1ccc(NC(=O)c2ccccc2NC(=O)c3ccc(cc3)[N+]([O-])=O)cc1

InChI

1S/C22H19N3O5/c26-14-13-15-5-9-17(10-6-15)23-22(28)19-3-1-2-4-20(19)24-21(27)16-7-11-18(12-8-16)25(29)30/h1-12,26H,13-14H2,(H,23,28)(H,24,27)

InChI key

LTWQQWSXYYXVGA-UHFFFAOYSA-N

Biochem/physiol Actions

KS-176 is a potent specific inhibitor of Breast Cancer Resistance Protein (BCRP) / ATP-binding cassette sub-family G member 2 (ABCG2) with no inhibition of the ATP binding cassette (ABC) transporters P-gp and MRP1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ronald E Cannon et al.
Toxicological sciences : an official journal of the Society of Toxicology, 169(2), 475-484 (2019-03-05)
Tetrabromobisphenol A (TBBPA, CAS No. 79-94-7) is a brominated flame retardant used in 90% of epoxy coated circuit boards. Exposures to TBBPA can induce neurotoxicity and disrupt MAPK, estrogen, thyroid, and PPAR-associated signaling pathways. Because these pathways also regulate transporters

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service