Skip to Content
Merck
All Photos(1)

Key Documents

SML1571

Sigma-Aldrich

S14161

≥98% (HPLC)

Synonym(s):

8-Ethoxy-2-(4-fluorophenyl)-3-nitro-2H-1-benzopyran, 8-Ethoxy-2-(4-fluorophenyl)-3-nitro-2H-chromene, Pichromene

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€64.90

€64.90


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€64.90

About This Item

Empirical Formula (Hill Notation):
C17H14FNO4
CAS Number:
Molecular Weight:
315.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€64.90


Estimated to ship on13 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

storage temp.

−20°C

SMILES string

FC(C=C1)=CC=C1C2C([N+]([O-])=O)=CC3=CC=CC(OCC)=C3O2

InChI

1S/C17H14FNO4/c1-2-22-15-5-3-4-12-10-14(19(20)21)16(23-17(12)15)11-6-8-13(18)9-7-11/h3-10,16H,2H2,1H3

InChI key

BFZXDHIUPNWOMT-UHFFFAOYSA-N

Biochem/physiol Actions

S14161 inhibits D-cyclin transactivation by inhibition of phosphoinositide 3-kinase (PI3K) activity. S14161 is a pan-class I PI3K inhibitor. S14161 inhibited the activity of all 4 isoforms of the PI3K class I family, and unlike LY294002, had little effect on class IV enzymes mechanistic target of rapamycin (mTOR) and DNA-PKcs (DNA-dependent protein kinase, catalytic subunit), or on AKT-1, -2, or -3 or PDK1. S14161 inhibited the expression of cyclins D1, D2, and D3, leading to the arrest of cells at the G0/G1 phase, and has exhibited anti-myeloma and anti-leukemia activity with no gross toxicity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo-Jiang Hou et al.
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association, 125(10), 669-676 (2017-09-13)
EXf, a glucagon-like peptide 1 (GLP-1) receptor agonist, stimulates β-cell proliferation and reduces apoptosis in diabetic animal models, but the underlying mechanisms are not fully understood. We constructed a FoxO1-GFP fusion protein expression plasmid and transiently transfected it into NIT-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service