Skip to Content
Merck
All Photos(1)

Key Documents

SML0463

Sigma-Aldrich

BPD

≥98% (HPLC)

Synonym(s):

3-(1,3-benzodioxol-5-yl)-4-phenyl-2,5-Furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H10O5
CAS Number:
Molecular Weight:
294.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 15 mg/mL (clear solution)

storage temp.

−20°C

SMILES string

O=C1OC(=O)C(c2ccc3OCOc3c2)=C1c4ccccc4

InChI

1S/C17H10O5/c18-16-14(10-4-2-1-3-5-10)15(17(19)22-16)11-6-7-12-13(8-11)21-9-20-12/h1-8H,9H2

InChI key

IMHGOAWBSJVILI-UHFFFAOYSA-N

Biochem/physiol Actions

BPD inhibits LPS-induced NF-kB signaling by inactivation of TAK-1. BPD is also known to inhibit the expression and activity of COX-2.
BPD inhibits LPS-induced NF-kB signaling; COX-2 inhibitor.

Features and Benefits

This compound is featured on the Cytokine Receptors (Interleukin-1 Receptor/TIR Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Karin Edstedt Bonamy et al.
Archives of disease in childhood. Fetal and neonatal edition, 104(1), F36-F45 (2018-01-22)
To investigate the variation in severe neonatal morbidity among very preterm (VPT) infants across European regions and whether morbidity rates are higher in regions with low compared with high mortality rates. Area-based cohort study of all births before 32 weeks
Sebastian Euler et al.
Frontiers in psychology, 9, 824-824 (2018-06-19)
Interpersonal sensitivity, particularly threat of potential exclusion, is a critical condition in borderline personality disorder (BPD) which impairs patients' social adjustment. Current evidence-based treatments include group components, such as mentalization-based group therapy (MBT-G), in order to improve interpersonal functioning. These
Jennifer Sophie Landry et al.
Annals of the American Thoracic Society, 13(1), 17-24 (2015-11-03)
Bronchopulmonary dysplasia and the long-term consequences of prematurity are underrecognized entities, unfamiliar to adult clinicians. Well described by the pediatric community, these young adults are joining the ranks of a growing population of adults with chronic lung disease. To describe
Johanna Köhling et al.
Journal of personality disorders, 30(6), 776-795 (2015-12-02)
The quality of depression in borderline personality disorder (BPD) was reported to differ from that in patients with major depressive disorder (MDD) only. However, little is known about affect dynamics in "borderline-depression." The authors assessed affective instability and reactivity in
Carla Sharp et al.
Assessment, 26(6), 1014-1029 (2018-07-14)
DSM-5 (Diagnostic and Statistical Manual of Mental Disorders, Fifth Edition) Section II criteria for borderline personality disorder (BPD) lack developmental operationalization. The aim of the current study was to evaluate whether DSM criteria operate similarly across adolescents and adults to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service