Skip to Content
Merck
All Photos(1)

Key Documents

SML0393

Sigma-Aldrich

QNZ46

≥98% (HPLC)

Synonym(s):

4-[6-Methoxy-2-[(1E)-2-(3-nitrophenyl)ethenyl]-4-oxo-3(4H)-quinazolinyl]-Benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H17N3O6
CAS Number:
Molecular Weight:
443.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL (warmed; clear solution)

storage temp.

room temp

SMILES string

COc1ccc2N=C(\C=C\c3cccc(c3)[N+]([O-])=O)N(C(=O)c2c1)c4ccc(cc4)C(O)=O

InChI

1S/C24H17N3O6/c1-33-19-10-11-21-20(14-19)23(28)26(17-8-6-16(7-9-17)24(29)30)22(25-21)12-5-15-3-2-4-18(13-15)27(31)32/h2-14H,1H3,(H,29,30)/b12-5+

InChI key

GNLVJIICVWDSNI-LFYBBSHMSA-N

Biochem/physiol Actions

QNZ46 is a noncompetitive inhibitor of GluN2C/D containing NMDA receptors. KD and IC50 values for binding and inhibition of GluN1/Glun2D receptors by QNZ46 are 4.9 and 3.9 μM, respectively. QNZ46 does not compete for binding of glutamate or glycine, but QNZ46 receptor binding requires the binding of glutamate to the GluN2 subunit.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reshmi Tognatta et al.
Frontiers in cellular neuroscience, 14, 74-74 (2020-04-18)
Astrocytes have been implicated in regulating oligodendrocyte development and myelination in vitro, although their functions in vivo remain less well defined. Using a novel approach to locally ablate GFAP+ astrocytes, we demonstrate that astrocytes are required for normal CNS myelin

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service