Skip to Content
Merck
All Photos(1)

Key Documents

SML3128

Sigma-Aldrich

HU-308

≥95% (HPLC)

Synonym(s):

(1S,4S,5S)-4-[4-(1,1-Dimethylheptyl)-2,6-dimethoxyphenyl]-6,6-dimethylbicyclo[3.1.1]hept-2-ene-2-methanol, HU308

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€127.00
25 MG
€509.00

€127.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€127.00
25 MG
€509.00

About This Item

Empirical Formula (Hill Notation):
C27H42O3
CAS Number:
Molecular Weight:
414.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€127.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

optical activity

[α]/D 180.0 to 130.0°, c = 0.5 in chloroform-d

storage condition

desiccated

color

white to dark beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−70°C

SMILES string

CCCCCCC(C)(C1=CC(OC)=C(C(OC)=C1)[C@]2(C=C([C@@H]3C(C)([C@H]2C3)C)CO)[H])C

InChI

1S/C27H42O3/c1-8-9-10-11-12-26(2,3)19-14-23(29-6)25(24(15-19)30-7)20-13-18(17-28)21-16-22(20)27(21,4)5/h13-15,20-22,28H,8-12,16-17H2,1-7H3/t20-,21+,22-/m0/s1

InChI key

CFMRIVODIXTERW-BDTNDASRSA-N

Biochem/physiol Actions

CB2 Cannabinoid receptor agonist
HU-308 is a potent, selective agonist of the cannabinoid receptor CB2. The Ki values for CB2 vs. CB1 are 22.7nM and >10 mM, respectively. HU-308 has anti-inflammatory and neuroprotective properties, and reduces liver damage in a hepatic ischemia/reperfusion injury model.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service