Skip to Content
Merck
All Photos(1)

Key Documents

SML0313

Sigma-Aldrich

Pimelic Diphenylamide 106

≥98% (HPLC)

Synonym(s):

Compound 106, Inhibitor 106, N1-(2-Aminophenyl)-N7-(4-methylphenyl)-heptanediamide, RGFA 8, TC-H 106

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30
25 MG
€286.00

€89.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€89.30
25 MG
€286.00

About This Item

Empirical Formula (Hill Notation):
C20H25N3O2
CAS Number:
Molecular Weight:
339.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€89.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ccc(NC(=O)CCCCCC(=O)Nc2ccccc2N)cc1

InChI

1S/C20H25N3O2/c1-15-11-13-16(14-12-15)22-19(24)9-3-2-4-10-20(25)23-18-8-6-5-7-17(18)21/h5-8,11-14H,2-4,9-10,21H2,1H3,(H,22,24)(H,23,25)

InChI key

WTKBRPXPNAKVEQ-UHFFFAOYSA-N

Biochem/physiol Actions

106 is a Class I HDAC inhibitor, demonstrating no activity against class II HDACs. It is a slow, tight-binding inhibitor of HDACs 1, 2, and 3, with a preference toward HDAC3 with Ki of 14 nM, 15 times lower than the Ki for HDAC1.
106 is a potent Class I HDAC inhibitor, demonstrating no activity against class II HDACs.
Pimelic diphenylamides has the ability to enhance the expression of the frataxin gene in lymphocytes from Friedreich ataxia patients.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pimelic diphenylamide 106 is a slow, tight-binding inhibitor of class I histone deacetylases
Chou CJ, et al.
The Journal of Biological Chemistry, 283(51), 35402-35409 (2008)
Yifei Liao et al.
PLoS pathogens, 17(2), e1009307-e1009307 (2021-02-18)
Marek's disease virus (MDV) is a potent oncogenic alphaherpesvirus that elicits a rapid onset of malignant T-cell lymphomas in chickens. Three MDV types, including GaHV-2 (MDV-1), GaHV-3 (MDV-2) and MeHV-1 (HVT), have been identified and all encode a US3 protein
Jason A Pfister et al.
PloS one, 14(4), e0215208-e0215208 (2019-04-12)
SIRT1, a NAD+-dependent deacetylase, protects neurons in a variety of in vitro and in vivo models of neurodegenerative disease. We have previously described a neuroprotective effect by SIRT1 independent of its catalytic activity. To confirm this conclusion we tested a
Dominik Stammler et al.
Journal of immunology (Baltimore, Md. : 1950), 195(11), 5421-5431 (2015-11-01)
Histone deacetylase (HDAC) inhibitors (HDACi) are clinically approved anticancer drugs that have important immune-modulatory properties. We report the surprising finding that HDACi promote LPS-induced IL-1β processing and secretion in human and murine dendritic cells and murine macrophages. HDACi/LPS-induced IL-1β maturation
Angela K Carrillo et al.
Bioorganic & medicinal chemistry, 23(16), 5151-5155 (2015-02-01)
Two of the histone deacetylases, TbDAC1 and TbDAC3, have been reported to be essential genes in trypanosomes. Therefore, we tested the activity of a panel of human histone deacetylase inhibitors (HDACi) for their ability to block proliferation of Trypanosoma brucei

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service