Skip to Content
Merck
All Photos(1)

Key Documents

O9390

Sigma-Aldrich

N-Oxalylglycine

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€99.70
50 MG
€395.00

€99.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€99.70
50 MG
€395.00

About This Item

Empirical Formula (Hill Notation):
C4H5NO5
CAS Number:
Molecular Weight:
147.09
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

€99.70


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

deionized water: >10 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)CNC(=O)C(O)=O

InChI

1S/C4H5NO5/c6-2(7)1-5-3(8)4(9)10/h1H2,(H,5,8)(H,6,7)(H,9,10)

InChI key

BIMZLRFONYSTPT-UHFFFAOYSA-N

Biochem/physiol Actions

N-Oxalylglycine is an inhibitor of α-ketoglutarate-dependent enzymes and mimics the initial steps but does not initiate the hydroxylation process. N-Oxalylglycine has been used to inhibit Jumonji C-domain-containing histone lysine demethylases.
N-Oxalylglycine is an inhibitor of α-ketoglutarate-dependent enzymes.

Features and Benefits

This compound is a featured product for Gene Regulation and Neuroscience research. Discover more featured Gene Regulation and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate/GABA Synthesis and Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gulzhan Raiymbek et al.
eLife, 9 (2020-03-21)
H3K9 methylation (H3K9me) specifies the establishment and maintenance of transcriptionally silent epigenetic states or heterochromatin. The enzymatic erasure of histone modifications is widely assumed to be the primary mechanism that reverses epigenetic silencing. Here, we reveal an inversion of this
Shan Shao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(8), 10860-10870 (2020-06-28)
The tumor microenvironment (TME) is a crucial factor in cancer progression. In breast cancer, cancer-associated fibroblasts (CAFs) and the derived stromal components have been recognized as comprising the majority of the pathological structure of the TME. In this study, we
Lisa P Elia et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(17), 3332-3344 (2019-01-31)
Deficient progranulin levels cause dose-dependent neurological syndromes: haploinsufficiency leads to frontotemporal lobar degeneration (FTLD) and nullizygosity produces adult-onset neuronal ceroid lipofuscinosis. Mechanisms controlling progranulin levels are largely unknown. To better understand progranulin regulation, we performed a genome-wide RNAi screen using
Nathaniel W Mabe et al.
Cell reports, 33(5), 108341-108341 (2020-11-05)
Dysregulated gene expression is a common feature of cancer and may underlie some aspects of tumor progression, including tumor relapse. Here, we show that recurrent mammary tumors exhibit global changes in gene expression and histone modifications and acquire dependence on
Xiongwen Cao et al.
Cell reports, 30(12), 4152-4164 (2020-03-27)
Histone methyl groups can be removed by demethylases. Although LSD1 and JmjC domain-containing proteins have been identified as histone demethylases, enzymes for many histone methylation states or sites are still unknown. Here, we perform a screening of a cDNA library

Articles

Glutamine's role in neurotransmitter synthesis and transport highlights its importance in neuronal function and glutamate production.

DISCOVER Bioactive Small Molecules for Neuroscience

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service