Skip to Content
Merck
All Photos(1)

Key Documents

SMB01037

Sigma-Aldrich

Robustaside D

≥90% (LC/MS-ELSD)

Synonym(s):

4-hydroxyphenyl 6-O-[(2E)-3-(1-hydroxy-4-oxocyclohexa-2,5-dien-1-yl)prop-2-enoyl]-beta-D-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€183.00

€183.00


Estimated to ship on21 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€183.00

About This Item

Empirical Formula (Hill Notation):
C21H22O10
CAS Number:
Molecular Weight:
434.39
UNSPSC Code:
12352205
NACRES:
NA.25

€183.00


Estimated to ship on21 April 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

434.39

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1COC(=O)\C=C\C3(C=CC(=O)C=C3)O)O)O)O)Oc2ccc(cc2)O

InChI

1S/C21H22O10/c22-12-1-3-14(4-2-12)30-20-19(27)18(26)17(25)15(31-20)11-29-16(24)7-10-21(28)8-5-13(23)6-9-21/h1-10,15,17-20,22,25-28H,11H2/b10-7+/t15-,17-,18+,19-,20-/m1/s1

InChI key

KQIQKULTIAJZKL-VHFDLOJPSA-N

General description

Robustaside D, a phenolic glycoside, is a bioactive natural product commonly derived from Grevillea robusta, Heliciopsis terminalis, and Helicia tsaii. Existing research suggests that this plant-derived metabolite exerts various biological activities, including antioxidant, hypoglycemic, and antimalarial properties.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon P B Ovenden et al.
Journal of natural products, 74(1), 74-78 (2010-12-16)
In search of new antimalarial compounds, three new phenolic glycosides, robustasides E (1), F (2), and G (3), in addition to the known compounds robustaside D (4) and quercetin-7-O-[α-l-rhamnopyranosyl(1→6)-β-d-galactopyranoside] (5), were identified during chemical investigations of the MeOH extract from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service