Skip to Content
Merck
All Photos(1)

Key Documents

SMB01030

Sigma-Aldrich

Theviridoside

≥90% (LC/MS-ELSD)

Synonym(s):

Methyl (1S,4aR,7aR)-1-(β-D-glucopyranosyloxy)-1,4a,5,7a-tetrahydro-4a-hydroxy-7-(hydroxymethyl)cyclopenta[c]pyran-4-carboxylate (ACI), Theveridoside, Theveside methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€204.00

€204.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€204.00

About This Item

Empirical Formula (Hill Notation):
C17H24O11
CAS Number:
Molecular Weight:
404.37
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

€204.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

404.37

solubility

water: soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2OC=C([C@@]3([C@H]2C(=CC3)CO)O)C(=O)OC

InChI

1S/C17H24O11/c1-25-14(23)8-6-26-15(10-7(4-18)2-3-17(8,10)24)28-16-13(22)12(21)11(20)9(5-19)27-16/h2,6,9-13,15-16,18-22,24H,3-5H2,1H3/t9-,10+,11-,12+,13-,15+,16+,17+/m1/s1

InChI key

LDBMLOLBWUOZGG-DOFVRBEMSA-N

General description

Theviridoside, an iridoid glycoside, is a natural product commonly found in plants of Cerbera odollam, Lippia javanica, Lippia turbinata and Thevetia neriifolia. In Cancer research, it exerts a significant effect on cell function and signal transduction, inducing cell cycle arrest and apoptosis in cancer cells while inhibiting the production of pro-inflammatory cytokines. This compound exhibits potent biological activity, including anti-cancer, antifungal, anti-inflammatory, and anti-viral activities.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Theviridoside acts by inhibiting the Na+/K+-ATPase pump, leading to increased intracellular calcium levels and the activation of multiple signaling pathways.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frédéric Martin et al.
Phytochemistry, 68(9), 1307-1311 (2007-03-27)
Sixteen crude extracts from six Panamanian plants of the family Bignoniaceae were submitted to rapid TLC tests against DPPH and acetylcholinesterase. Pithecoctenium crucigerum (L.) A.H. Gentry, which showed interesting activity against DPPH, has been studied. The chemical investigation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service