Skip to Content
Merck
All Photos(1)

Key Documents

SMB01076

Sigma-Aldrich

Glycycoumarin

≥90% (LC/MS-ELSD)

Synonym(s):

3-(2,4-dihydroxyphenyl)-7-hydroxy-5-methoxy-6-(3-methylbut-2-enyl)chromen-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€306.00

€306.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€306.00

About This Item

Empirical Formula (Hill Notation):
C21H20O6
CAS Number:
Molecular Weight:
368.38
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

€306.00


Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

368.38

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[o]1c2c(c(c(c(c2)O)CC=C(C)C)OC)cc([c]1=O)c3c(cc(cc3)O)O

InChI

1S/C21H20O6/c1-11(2)4-6-14-18(24)10-19-16(20(14)26-3)9-15(21(25)27-19)13-7-5-12(22)8-17(13)23/h4-5,7-10,22-24H,6H2,1-3H3

InChI key

NZYSZZDSYIBYLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycycoumarin, a major bioactive coumarin, is an aromatic ether and a member of resorcinols. This bioactive natural product is commonly derived from Glycyrrhiza Sp. (G. glabra, G. uralensis and G. radix) plants. Existing research suggests that this plant-derived metabolite exerts various biological activities, including anti-spasmodic, hepatoprotective, anticancer, neuroprotective, antioxidant and anti-inflammatory activities.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

According to the existing research, Glycycoumarin serves as an estrogen agonist while also exhibiting moderate inhibitory effects on enzymes CYP1A2 and CYP2B6. Notably, Glycycoumarin demonstrates potent anti-hepatitis C virus (HCV) activity, with an IC(50) value of 8.8 µg/mL. Furthermore, it effectively inhibits smooth muscle contraction induced by various stimuli by targeting PDEs, especially isozyme 3, which leads to the accumulation of intracellular cAMP, a vital component of cellular signaling. Additionally, Glycycoumarin acts as a potent antispasmodic through the inhibition of phosphodiesterase 3, further underscoring its significance in modulating cellular responses and health. Moreover, it plays a crucial role in inhibiting hepatocyte lipoapoptosis by activating autophagy and inhibiting the ER stress/GSK-3-mediated mitochondrial pathway, showcasing its importance in cellular function.
Glycycoumarin exhibits antispasmodic activity and has a role as an antispasmodic drug and a plant metabolite. It has been demostrated to be a novel active component of licorice against non-alcoholic fatty liver disease (NAFLD).

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service