Skip to Content
Merck
All Photos(1)

Key Documents

SMB00280

Sigma-Aldrich

D-Mannuronic acid sodium

≥90% (HPLC)

Synonym(s):

D-Mannuronic acid sodium, Mono-mannuronic acid sodium salt, Sodium mannuronate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€350.00

€350.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€350.00

About This Item

Empirical Formula (Hill Notation):
C6H9NaO7
CAS Number:
Molecular Weight:
216.12
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.28

€350.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white to off-white

solubility

water: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

O[C@H]([C@@H](C(O[Na])=O)O[C@@H](O)[C@H]1O)[C@@H]1O

InChI

1S/C6H10O7.Na/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;/h1-4,6-9,12H,(H,10,11);/q;+1/p-1/t1?,2-,3+,4?,6+;/m0./s1

InChI key

MSXHSNHNTORCAW-NMRVOGSPSA-M

Application


  • Effects of β-d-mannuronic acid, as a novel non-steroidal anti-inflammatory medication with immunosuppressive properties, on IL17, RORγt, IL4, and GATA3 gene expressions in rheumatoid arthritis patients.: This research examines the impact of β-d-mannuronic acid on gene expressions related to inflammation and immune responses in rheumatoid arthritis, suggesting its use as an anti-inflammatory drug (Barati et al., 2017).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magda Dudek et al.
Nucleic acids research, 48(14), 7786-7800 (2020-06-26)
Marine flavobacteria possess dedicated Polysaccharide Utilization Loci (PULs) enabling efficient degradation of a variety of algal polysaccharides. The expression of these PULs is tightly controlled by the presence of the substrate, yet details on the regulatory mechanisms are still lacking.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service