Skip to Content
Merck
All Photos(1)

Key Documents

G1951

Sigma-Aldrich

D-Gluconic acid solution

49-53 wt. % in H2O

Synonym(s):

2,3,4,5,6-Pentahydroxycaproic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€25.30
1 KG
€56.60
4 KG
€144.00

€25.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€25.30
1 KG
€56.60
4 KG
€144.00

About This Item

Empirical Formula (Hill Notation):
C6H12O7
CAS Number:
Molecular Weight:
196.16
Beilstein:
1726055
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

€25.30


Check Cart for Availability

Request a Bulk Order

form

viscous liquid

Quality Level

optical activity

[α]/D +9.0 to 15.5°

concentration

49-53 wt. % in H2O

refractive index

n20/D 1.4161

density

1.234 g/mL at 25 °C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O

InChI

1S/C6H12O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h2-5,7-11H,1H2,(H,12,13)/t2-,3-,4+,5-/m1/s1

InChI key

RGHNJXZEOKUKBD-SQOUGZDYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Gluconic acid is a non-volatile, organic acid generally used as a sequestering agent and as a scale inhibitor.[1] Due to its tendency to chelate with earth alkaline elements and heavy metals, it is also used as a metal extractant.[1] Gluconic acid and its salts are
used in hygienic products, food and pharmaceutical industry.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of heavy metals from soil components and soils by natural chelating agents. Part II. Soil extraction by sugar acids.
Fischer K and Bipp HP
Water, Air, and Soil Pollution, 138(1-4), 271-288 (2002)
Gluconic acid: Properties, applications and microbial production.
Ramachandran S, et al.
Food Technology and Biotechnology, 44(2) (2006)
Sheba Khan et al.
PloS one, 15(8), e0238252-e0238252 (2020-08-29)
We report on aerobic "environmental" bacteria isolated from European honey bees (Apis mellifera). We determined the number of culturable aerobic bacteria in the gut of nurse bees sampled from locations around Australia. Bees from healthy colonies had 107-108 aerobic bacteria
Mallory L Hammock et al.
Advanced materials (Deerfield Beach, Fla.), 26(35), 6138-6144 (2014-07-23)
Organic field-effect transistor (OFET) sensors can meet the need for portable and real-time diagnostics. An electronicreadout enzyme-linked immunosorbent assay using OFETs for the detection of a panel of three biomarkers in complex media to create a pre-eclampsia prognostic is demonstrated
Ubaid H Shah et al.
Clinical nutrition (Edinburgh, Scotland), 32(2), 193-199 (2012-09-18)
Acute lower respiratory infections are the most frequent illnesses globally in children less than 5 years old. The aim of this randomized double blind controlled trial is to assess the effectiveness of zinc gluconate supplementation for 2 months period compared

Questions

1–5 of 5 Questions  
  1. Does this product contain potassium or sodium as a component?

    1 answer
    1. This product is prepared from the Gluconic acid and not a potassium or sodium salt form. While trace analysis testing is not performed on this material, no potassium or sodium has been added during the manufacturing process.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  4. WHAT is the ppm of the solution in 100ml

    1 answer
    1. The concentration of this product may range from 49% - 53%. The calculated ppm value will range from 490,000 - 530,000.

      Helpful?

  5. Hi what is the molarity of the solution provided? 25g in 50%wt solution

    1 answer
    1. At exactly 50% the molarity of this solution is 3.14M. The molarity may be calculated using the following equation below:

      Molarity (moles/L) = Density (g/L) ÷ Molecular Weight (g/mol) * purity
      = 1234g/L ÷ 196.16 g/mol * 0.50
      = 3.14539M

      The concentration of this product ranges from 49 - 53% and may change slightly from lot to lot. Please see the link below to access a sample or specific lot Certificate of Analysis:
      https://www.sigmaaldrich.com/product/aldrich/g1951#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service